Orders LinkContactUsLinkComponent
Masterflex® Variable Area Flowmeter Multitube Frames, Avantor®
Masterflex® Variable Area Flowmeter Multitube Frames, Avantor®
Masterflex® Variable Area Flowmeter Multitube Frames, Avantor®
Certificates
Masterflex® Variable Area Flowmeter Multitube Frames, Avantor®
Masterflex® Variable Area Flowmeter Multitube Frames, Avantor®
37 Options Available Below
Product Details & Documents
Create your own flowmeter systems for complete flexibility.
  • Correlated flowmeters feature a generic scale—reference flow curves calibrated to a variety of media, temperatures and pressures to get the flow rate
  • Direct-reading flowmeters feature a calibrated scale for a specific medium—no need to convert readings to get flow rate
  • Use with media for laboratory and industrial applications
  • Required system components for flowmeter systems would be frame, flowtube(s), valve cartridge, and tripod—order each component separately
  • Choose from aluminum or stainless steel wetted parts based on required chemical compatibility
  • Designed with 1/8” NPT(F) connections
Design a flow system to meet your metering, mixing, or monitoring applications. We offer a variety of frames and flowtubes so that you can create the system to meet your exact needs.
Choose a frame depending on the flowtube capacity, flow pattern, and wetted parts that you require.
These frames have valve ports which accept valve cartridges to control flow. A tripod base is required to use the multitube frame with the common inlet flow pattern; all other multitube frames are designed for panel and tripod mounting.
Specifications for Product Family
Specifications
  • Max. operating temperature:
  • 121 °C (250 °F)
  • 121 °C (250 °F)
  • 121 °C (250 °F)
  • 121 °C (250 °F)
  • 121 °C (250 °F)
  • 121 °C (250 °F)
  • 121 °C (250 °F)
  • 121 °C (250 °F)
  • 121 °C (250 °F)
  • 121 °C (250 °F)
  • 121 °C (250 °F)
  • 121 °C (250 °F)
  • 121 °C (250 °F)
  • 121 °C (250 °F)
  • 121 °C (250 °F)
  • 121 °C (250 °F)
  • 121 °C (250 °F)
  • 121 °C (250 °F)
  • 121 °C (250 °F)
  • 121 °C (250 °F)
  • 121 °C (250 °F)
  • 121 °C (250 °F)
  • 121 °C (250 °F)
  • 121 °C (250 °F)
  • 121 °C (250 °F)
  • 121 °C (250 °F)
  • 121 °C (250 °F)
  • 121 °C (250 °F)
  • 121 °C (250 °F)
  • 121 °C (250 °F)
  • 121 °C (250 °F)
  • 121 °C (250 °F)
  • 121 °C (250 °F)
  • 121 °C (250 °F)
  • 121 °C (250 °F)
  • 121 °C (250 °F)
  • 121 °C (250 °F)
  • Warranty:
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
Specifications
Product Family Options
Product Information
  • Process connection
    Availability
    Price
  • MFLX03214-10
    Two Tube Frame for 65 mm Flowtube, Isolated, Aluminum
    Two Tube Frame for 65 mm Flowtube, Isolated, Aluminum
    0,125" NPT(F)
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 158,00 €
    158,00 €
    1 * 1 ST
       158,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    MFLX03214-10
    Max. operating temperature
    121 °C (250 °F)
    Warranty
    1 year
  • MFLX03214-12
    Two Tube Frame for 65 mm Flowtube, Common, Aluminum
    Two Tube Frame for 65 mm Flowtube, Common, Aluminum
    0,125" NPT(F)
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 195,00 €
    195,00 €
    1 * 1 ST
       195,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    MFLX03214-12
    Max. operating temperature
    121 °C (250 °F)
    Warranty
    1 year
  • MFLX03214-16
    Two Tube Frame for 65 mm Flowtube, Isolated, 316 SS
    Two Tube Frame for 65 mm Flowtube, Isolated, 316 SS
    0,125" NPT(F)
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 407,00 €
    407,00 €
    1 * 1 ST
       407,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    MFLX03214-16
    Max. operating temperature
    121 °C (250 °F)
    Warranty
    1 year
  • MFLX03214-18
    Two Tube Frame for 65 mm Flowtube, Common, 316 SS
    Two Tube Frame for 65 mm Flowtube, Common, 316 SS
    0,125" NPT(F)
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 432,00 €
    432,00 €
    1 * 1 ST
       432,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    MFLX03214-18
    Max. operating temperature
    121 °C (250 °F)
    Warranty
    1 year
  • MFLX03214-28
    Three Tube Frame for 65 mm Flowtube, Isolated, Aluminum
    Three Tube Frame for 65 mm Flowtube, Isolated, Aluminum
    0,125" NPT(F)
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 197,00 €
    197,00 €
    1 * 1 ST
       197,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    MFLX03214-28
    Max. operating temperature
    121 °C (250 °F)
    Warranty
    1 year
  • MFLX03214-30
    Three Tube Frame for 65 mm Flowtube, Common, Aluminum
    Three Tube Frame for 65 mm Flowtube, Common, Aluminum
    0,125" NPT(F)
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 240,00 €
    240,00 €
    1 * 1 ST
       240,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    MFLX03214-30
    Max. operating temperature
    121 °C (250 °F)
    Warranty
    1 year
  • MFLX03214-36
    Three Tube Frame for 65 mm Flowtube, Common, 316 SS
    Three Tube Frame for 65 mm Flowtube, Common, 316 SS
    0,125" NPT(F)
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 616,00 €
    616,00 €
    1 * 1 ST
       616,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    MFLX03214-36
    Max. operating temperature
    121 °C (250 °F)
    Warranty
    1 year
  • MFLX03214-46
    Four Tube Frame for 65 mm Flowtube, Isolated, Aluminum
    Four Tube Frame for 65 mm Flowtube, Isolated, Aluminum
    0,125" NPT(F)
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 257,00 €
    257,00 €
    1 * 1 ST
       257,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    MFLX03214-46
    Max. operating temperature
    121 °C (250 °F)
    Warranty
    1 year
  • MFLX03214-50
    Four Tube Frame for 65 mm Flowtube, Common, Aluminum
    Four Tube Frame for 65 mm Flowtube, Common, Aluminum
    0,125" NPT(F)
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 297,00 €
    297,00 €
    1 * 1 ST
       297,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    MFLX03214-50
    Max. operating temperature
    121 °C (250 °F)
    Warranty
    1 year
  • MFLX03214-52
    Four Tube Frame for 65 mm Flowtube, Isolated, 316 SS
    Four Tube Frame for 65 mm Flowtube, Isolated, 316 SS
    0,125" NPT(F)
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 757,00 €
    757,00 €
    1 * 1 ST
       757,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    MFLX03214-52
    Max. operating temperature
    121 °C (250 °F)
    Warranty
    1 year
  • MFLX03214-56
    Four Tube Frame for 65 mm Flowtube, Common, 316 SS
    Four Tube Frame for 65 mm Flowtube, Common, 316 SS
    0,125" NPT(F)
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 809,00 €
    809,00 €
    1 * 1 ST
       809,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    MFLX03214-56
    Max. operating temperature
    121 °C (250 °F)
    Warranty
    1 year
  • MFLX03214-62
    Five Tube Frame for 65 mm Flowtube, Isolated, Aluminum
    Five Tube Frame for 65 mm Flowtube, Isolated, Aluminum
    0,125" NPT(F)
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 315,00 €
    315,00 €
    1 * 1 ST
       315,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    MFLX03214-62
    Max. operating temperature
    121 °C (250 °F)
    Warranty
    1 year
  • MFLX03214-64
    Five Tube Frame for 65 mm Flowtube, Common, Aluminum
    Five Tube Frame for 65 mm Flowtube, Common, Aluminum
    0,125" NPT(F)
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 364,00 €
    364,00 €
    1 * 1 ST
       364,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    MFLX03214-64
    Max. operating temperature
    121 °C (250 °F)
    Warranty
    1 year
  • MFLX03214-66
    Five Tube Frame for 65 mm Flowtube, Isolated, 316 SS
    Five Tube Frame for 65 mm Flowtube, Isolated, 316 SS
    0,125" NPT(F)
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 934,00 €
    934,00 €
    1 * 1 ST
       934,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    MFLX03214-66
    Max. operating temperature
    121 °C (250 °F)
    Warranty
    1 year
  • MFLX03214-68
    Five Tube Frame for 65 mm Flowtube, Common, 316 SS
    Five Tube Frame for 65 mm Flowtube, Common, 316 SS
    0,125" NPT(F)
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 990,00 €
    990,00 €
    1 * 1 ST
       990,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    MFLX03214-68
    Max. operating temperature
    121 °C (250 °F)
    Warranty
    1 year
  • MFLX03214-74
    Six Tube Frame for 65 mm Flowtube, Isolated, Aluminum
    Six Tube Frame for 65 mm Flowtube, Isolated, Aluminum
    0,125" NPT(F)
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 363,00 €
    363,00 €
    1 * 1 ST
       363,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    MFLX03214-74
    Max. operating temperature
    121 °C (250 °F)
    Warranty
    1 year
  • MFLX03214-76
    Six Tube Frame for 65 mm Flowtube, Common, Aluminum
    Six Tube Frame for 65 mm Flowtube, Common, Aluminum
    0,125" NPT(F)
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 429,00 €
    429,00 €
    1 * 1 ST
       429,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    MFLX03214-76
    Max. operating temperature
    121 °C (250 °F)
    Warranty
    1 year
  • MFLX03214-78
    Six Tube Frame for 65 mm Flowtube, Isolated, 316 SS
    Six Tube Frame for 65 mm Flowtube, Isolated, 316 SS
    0,125" NPT(F)
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 130,00 €
    1 130,00 €
    1 * 1 ST
       1 130,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    MFLX03214-78
    Max. operating temperature
    121 °C (250 °F)
    Warranty
    1 year
  • MFLX03214-80
    Six Tube Frame for 65 mm Flowtube, Common, 316 SS
    Six Tube Frame for 65 mm Flowtube, Common, 316 SS
    0,125" NPT(F)
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 200,00 €
    1 200,00 €
    1 * 1 ST
       1 200,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    MFLX03214-80
    Max. operating temperature
    121 °C (250 °F)
    Warranty
    1 year
  • MFLX03215-10
    Two Tube Frame for 150 mm Flowtube, Isolated, Aluminum
    Two Tube Frame for 150 mm Flowtube, Isolated, Aluminum
    0,125" NPT(F)
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 224,00 €
    224,00 €
    1 * 1 ST
       224,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    MFLX03215-10
    Max. operating temperature
    121 °C (250 °F)
    Warranty
    1 year
  • MFLX03215-12
    Two Tube Frame for 150 mm Flowtube, Common, Aluminum
    Two Tube Frame for 150 mm Flowtube, Common, Aluminum
    0,125" NPT(F)
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 258,00 €
    258,00 €
    1 * 1 ST
       258,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    MFLX03215-12
    Max. operating temperature
    121 °C (250 °F)
    Warranty
    1 year
  • MFLX03215-16
    Two Tube Frame for 150 mm Flowtube, Isolated, 316 SS
    Two Tube Frame for 150 mm Flowtube, Isolated, 316 SS
    0,125" NPT(F)
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 453,00 €
    453,00 €
    1 * 1 ST
       453,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    MFLX03215-16
    Max. operating temperature
    121 °C (250 °F)
    Warranty
    1 year
  • MFLX03215-18
    Two Tube Frame for 150 mm Flowtube, Common, 316 SS
    Two Tube Frame for 150 mm Flowtube, Common, 316 SS
    0,125" NPT(F)
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 485,00 €
    485,00 €
    1 * 1 ST
       485,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    MFLX03215-18
    Max. operating temperature
    121 °C (250 °F)
    Warranty
    1 year
  • MFLX03215-28
    Three Tube Frame for 150 mm Flowtube, Isolated, Aluminum
    Three Tube Frame for 150 mm Flowtube, Isolated, Aluminum
    0,125" NPT(F)
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 288,00 €
    288,00 €
    1 * 1 ST
       288,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    MFLX03215-28
    Max. operating temperature
    121 °C (250 °F)
    Warranty
    1 year
  • MFLX03215-30
    Three Tube Frame for 150 mm Flowtube, Common, Aluminum
    Three Tube Frame for 150 mm Flowtube, Common, Aluminum
    0,125" NPT(F)
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 329,00 €
    329,00 €
    1 * 1 ST
       329,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    MFLX03215-30
    Max. operating temperature
    121 °C (250 °F)
    Warranty
    1 year
  • MFLX03215-34
    Three Tube Frame for 150 mm Flowtube, Isolated, 316 SS
    Three Tube Frame for 150 mm Flowtube, Isolated, 316 SS
    0,125" NPT(F)
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 662,00 €
    662,00 €
    1 * 1 ST
       662,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    MFLX03215-34
    Max. operating temperature
    121 °C (250 °F)
    Warranty
    1 year
  • MFLX03215-36
    Three Tube Frame for 150 mm Flowtube, Common, 316 SS
    Three Tube Frame for 150 mm Flowtube, Common, 316 SS
    0,125" NPT(F)
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 707,00 €
    707,00 €
    1 * 1 ST
       707,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    MFLX03215-36
    Max. operating temperature
    121 °C (250 °F)
    Warranty
    1 year
  • MFLX03215-46
    Four Tube Frame for 150 mm Flowtube, Isolated, Aluminum
    Four Tube Frame for 150 mm Flowtube, Isolated, Aluminum
    0,125" NPT(F)
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 376,00 €
    376,00 €
    1 * 1 ST
       376,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    MFLX03215-46
    Max. operating temperature
    121 °C (250 °F)
    Warranty
    1 year
  • MFLX03215-50
    Four Tube Frame for 150 mm Flowtube, Common, Aluminum
    Four Tube Frame for 150 mm Flowtube, Common, Aluminum
    0,125" NPT(F)
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 416,00 €
    416,00 €
    1 * 1 ST
       416,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    MFLX03215-50
    Max. operating temperature
    121 °C (250 °F)
    Warranty
    1 year
  • MFLX03215-52
    Four Tube Frame for 150 mm Flowtube, Isolated, 316 SS
    Four Tube Frame for 150 mm Flowtube, Isolated, 316 SS
    0,125" NPT(F)
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 871,00 €
    871,00 €
    1 * 1 ST
       871,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    MFLX03215-52
    Max. operating temperature
    121 °C (250 °F)
    Warranty
    1 year
  • MFLX03215-56
    Four Tube Frame for 150 mm Flowtube, Common, 316 SS
    Four Tube Frame for 150 mm Flowtube, Common, 316 SS
    0,125" NPT(F)
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 923,00 €
    923,00 €
    1 * 1 ST
       923,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    MFLX03215-56
    Max. operating temperature
    121 °C (250 °F)
    Warranty
    1 year
  • MFLX03215-64
    Five Tube Frame for 150 mm Flowtube, Common, Aluminum
    Five Tube Frame for 150 mm Flowtube, Common, Aluminum
    0,125" NPT(F)
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 428,00 €
    428,00 €
    1 * 1 ST
       428,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    MFLX03215-64
    Max. operating temperature
    121 °C (250 °F)
    Warranty
    1 year
  • MFLX03215-68
    Five Tube Frame for 150 mm Flowtube, Common, 316 SS
    Five Tube Frame for 150 mm Flowtube, Common, 316 SS
    0,125" NPT(F)
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 150,00 €
    1 150,00 €
    1 * 1 ST
       1 150,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    MFLX03215-68
    Max. operating temperature
    121 °C (250 °F)
    Warranty
    1 year
  • MFLX03215-74
    Six Tube Frame for 150 mm Flowtube, Isolated, Aluminum
    Six Tube Frame for 150 mm Flowtube, Isolated, Aluminum
    0,125" NPT(F)
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 534,00 €
    534,00 €
    1 * 1 ST
       534,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    MFLX03215-74
    Max. operating temperature
    121 °C (250 °F)
    Warranty
    1 year
  • MFLX03215-76
    Six Tube Frame for 150 mm Flowtube, Common, Aluminum
    Six Tube Frame for 150 mm Flowtube, Common, Aluminum
    0,125" NPT(F)
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 599,00 €
    599,00 €
    1 * 1 ST
       599,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    MFLX03215-76
    Max. operating temperature
    121 °C (250 °F)
    Warranty
    1 year
  • MFLX03215-78
    Six Tube Frame for 150 mm Flowtube, Isolated, 316 SS
    Six Tube Frame for 150 mm Flowtube, Isolated, 316 SS
    0,125" NPT(F)
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 310,00 €
    1 310,00 €
    1 * 1 ST
       1 310,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    MFLX03215-78
    Max. operating temperature
    121 °C (250 °F)
    Warranty
    1 year
  • MFLX03215-80
    Six Tube Frame for 150 mm Flowtube, Common, 316 SS
    Six Tube Frame for 150 mm Flowtube, Common, 316 SS
    0,125" NPT(F)
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 370,00 €
    1 370,00 €
    1 * 1 ST
       1 370,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    MFLX03215-80
    Max. operating temperature
    121 °C (250 °F)
    Warranty
    1 year