ContactUsLinkComponent
Masterflex® Pressure Booster Pump Systems, Avantor®
Masterflex® Pressure Booster Pump Systems, Avantor®
Certificates
undefined
Masterflex® Pressure Booster Pump Systems, Avantor®
Specifications
  • Model:
  • 70794-64
  • 70794-44
  • Max. pressure:
  • 3,9 bar (56 psi)
  • 4,3 bar (63 psi)
  • Motor enclosure type:
  • NEMA 4
  • NEMA 4
  • Motor hp:
  • 3
  • 2
  • Wetted materials:
  • SS pump, brass piping/valves/drain
  • SS pump, brass piping/valves/drain
  • Warranty:
  • 1 year
  • 1 year