ContactUsLinkComponent
Valved Acrylic Flowmeters for Bench or Panel Mount
Valved Acrylic Flowmeters for Bench or Panel Mount
Certificates
undefined
Valved Acrylic Flowmeters for Bench or Panel Mount
Specifications
  • Accuracy:
  • ±3% full-scale
  • ±3% full-scale
  • ±2% full-scale
  • ±3% full-scale
  • ±2% full-scale
  • ±3% full-scale
  • ±3% full-scale
  • ±3% full-scale
  • ±3% full-scale
  • ±3% full-scale
  • Certification:
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Float material:
  • 316 SS
  • 316 SS
  • 316 SS
  • 316 SS
  • 316 SS
  • 316 SS
  • 316 SS
  • 316 SS
  • 316 SS
  • 316 SS
  • Max. operating temperature:
  • 65 °C (150 °F)
  • 65 °C (150 °F)
  • 65 °C (150 °F)
  • 65 °C (150 °F)
  • 65 °C (150 °F)
  • 65 °C (150 °F)
  • 65 °C (150 °F)
  • 65 °C (150 °F)
  • 65 °C (150 °F)
  • 65 °C (150 °F)
  • O-Ring material:
  • Buna-N
  • Buna-N
  • Buna-N
  • Buna-N
  • Buna-N
  • Buna-N
  • Buna-N
  • Buna-N
  • Buna-N
  • Buna-N
  • Repeatability:
  • ±0,5% full-scale
  • ±0,5% full-scale
  • ±0,5% full-scale
  • ±0,5% full-scale
  • ±0,5% full-scale
  • ±0,5% full-scale
  • ±0,5% full-scale
  • ±0,5% full-scale
  • ±0,5% full-scale
  • ±0,5% full-scale
  • Warranty:
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year