ContactUsLinkComponent
Submersible Pumps for Corrosive/Acid Sumps
Submersible Pumps for Corrosive/Acid Sumps
Certificates
undefined
Submersible Pumps for Corrosive/Acid Sumps
Specifications
  • Duty cycle continuous:
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Max. pressure:
  • 8,6 bar (125 psi)
  • 8,6 bar (125 psi)
  • 8,6 bar (125 psi)
  • 8,6 bar (125 psi)
  • 8,6 bar (125 psi)
  • Max. temperature:
  • 57 °C (135 °F)
  • 57 °C (135 °F)
  • 57 °C (135 °F)
  • 57 °C (135 °F)
  • 57 °C (135 °F)
  • Motor type:
  • Manual
  • Manual
  • Automatic
  • Automatic
  • Manual
  • Motor hp:
  • 0,4
  • 0,4
  • 0,4
  • 0,4
  • 0,4
  • Outlet connection:
  • 1,25" NPT(F) inside thread; 1,5" tapered hose connection
  • 1,25" NPT(F) inside thread; 1,5" tapered hose connection
  • 1,25" NPT(F) inside thread; 1,5" tapered hose connection
  • 1,25" NPT(F) inside thread; 1,5" tapered hose connection
  • 1,25" NPT(F) inside thread; 1,5" tapered hose connection
  • Wetted materials:
  • Noryl/316 SS rotor shaft & hardware, Viton® seals
  • Noryl/titanium shaft & hardware, Viton® seals
  • Noryl/304 SS rotor shaft, 316 SS hardware, Viton®/Buna seals
  • Noryl/316 SS rotor shaft & hardware, Viton® seals
  • Noryl/304 SS rotor shaft, 316 SS hardware, Viton®/Buna seals
  • Warranty:
  • 3 Months
  • 3 Months
  • 3 Months
  • 3 Months
  • 3 Months