To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Length:22,5 cm (8,875")
- Max. capacity:48 ml
- Max. speed:15000 rpm
- Noise level:<51 dB(A)
- Power:120 VAC, 50/60 Hz
- Description:Ventilated Microcentrifuge with 24-tube rotor - 120V, 50/60 Hz
- Height with lid open:44 cm (17")
- Rotor type:Fixed-Angle
- Speed control:Touch Pad
- Tube capacity:24 x 1,5/2,0 mL
- Width:23,4 cm (9,2031")
- Max. RCF:21130 ×g
- Height:32 cm (12,625")
- Warranty:2 years
- Pk:1
- Rotors included:Yes
Added versatility to spin microcentrifuge tubes, miniprep spin columns, and PCR tube strips up to 15,000 rpm.
- Lid automatically opens at end of run for easy sample removal
- Suitable for use in cold rooms
- Whisper-quiet operation
Unit features microprocessor-based control with five-digit speed display of time and speed/g force. Easy-to-access control options include short-spin speed setting, soft-braking, “At Set” timer function, and adjustable volume alarm. Smooth keypad provides easy wipe-down cleaning. Aerosol-tight rotor is suitable even for hazardous materials, and contoured bowl design minimizes noise. Order optional rotors, below right, for added versatility.
- Length:22,5 cm (8,875")
- Max. capacity:48 ml
- Max. speed:15000 rpm
- Noise level:<51 dB(A)
- Power:120 VAC, 50/60 Hz
- Description:Ventilated Microcentrifuge with 24-tube rotor - 120V, 50/60 Hz
- Environmentally Preferable:
- Height with lid open:44 cm (17")
- Rotor type:Fixed-Angle
- Speed control:Touch Pad
- Tube capacity:24 x 1,5/2,0 mL
- Width:23,4 cm (9,2031")
- Max. RCF:21130 ×g
- Height:32 cm (12,625")
- Warranty:2 years
- Pk:1
- Rotors included:Yes