ContactUsLinkComponent
Signet DIN Flow Monitors and Controllers
GF Signet 3-8150-1 8150 Field Mount Flow Monitor/Totalizer, Battery Powered
Catalog # MFLX32487-29
undefined
GF Signet 3-8150-1 8150 Field Mount Flow Monitor/Totalizer, Battery Powered
Catalog # MFLX32487-29
Specifications
  • Description:
    8150 Field Mount Flow Monitor/Totalizer, Battery Powered
  • Warranty:
    1 year
  • Pk:
    1
Specifications
About this item
Easy read your flow.
  • Loop-Powered Digital Flow Transmitter
  • Battery-Powered Digital Flow Rate Monitor and Totalizer
  • Line-Powered Analog/Digital Flow Controller
  • Batch Controller
This field mount flow monitor/totalizer is battery powered for optimal convience.
  • Description:
    8150 Field Mount Flow Monitor/Totalizer, Battery Powered
  • Environmentally Preferable:
  • Warranty:
    1 year
  • Pk:
    1