ContactUsLinkComponent
Male Cam to Male NPT Threaded Adapters, Polypropylene
Cam Fitting, Polypropylene, Male Cam x 3/4" NPT(M)
Catalog # MFLX34020-02
undefined
Cam Fitting, Polypropylene, Male Cam x 3/4" NPT(M)
Catalog # MFLX34020-02
Specifications
  • Connection Type:
    Cam to Thread
  • Flow pattern:
    Straight
  • Thread size NPT(M) (port 1):
    ¾"
  • Seal material:
    EPDM
  • Description:
    Cam Fitting, Polypropylene, Male Cam×¾" NPT(M)
  • Warranty:
    1 Month
  • Pk:
    1
  • Fitting material:
    Polypropylene
Specifications
About this item
Cam couplers provide quick disconnection from system.
  • All fittings have EPDM gaskets
  • Constructed of durable polypropylene
Cam couplers are ideal for providing a quick-disconnect solution to your pump line.
  • Connection Type:
    Cam to Thread
  • Flow pattern:
    Straight
  • Thread size NPT(M) (port 1):
    ¾"
  • Seal material:
    EPDM
  • Description:
    Cam Fitting, Polypropylene, Male Cam×¾" NPT(M)
  • Environmentally Preferable:
  • Warranty:
    1 Month
  • Pk:
    1
  • Fitting material:
    Polypropylene