Cam Fitting, Polypropylene, Female Cam x 3/4" NPT(M)
Catalog # MFLX34020-32
Supplier: Avantor Fluid Handling
Specifications
- Connection Type:Cam to Thread
- Flow pattern:Straight
- Thread size NPT(M) (port 1):¾"
- Seal material:EPDM
- Description:Cam Fitting, Polypropylene, Female Cam×¾" NPT(M)
- Warranty:1 Month
- Pk:1
- Fitting material:Polypropylene
Specifications
About this item
Cam couplers provide quick disconnection from system.
- All fittings have EPDM gaskets
- Constructed of durable polypropylene
Cam couplers are ideal for providing a quick-disconnect solution to your pump line.
- Connection Type:Cam to Thread
- Flow pattern:Straight
- Thread size NPT(M) (port 1):¾"
- Seal material:EPDM
- Description:Cam Fitting, Polypropylene, Female Cam×¾" NPT(M)
- Environmentally Preferable:
- Warranty:1 Month
- Pk:1
- Fitting material:Polypropylene