Cam Fitting, Polypropylene, Female Cam x 1-1/2" Hose Barb
Catalog # MFLX34020-51
Supplier: Avantor Fluid Handling
Specifications
- Barb size/tubing int.Ø (port 1):1½"
- Connection Type:Cam to Barb
- Flow pattern:Straight
- Seal material:EPDM
- Description:Cam Fitting, Polypropylene, Female Cam×1½" Hose Barb
- Warranty:1 Month
- Pk:1
- Fitting material:Polypropylene
Specifications
About this item
Cam couplers provide quick disconnection from system.
- All fittings have EPDM gaskets
- Constructed of durable polypropylene
Cam couplers are ideal for providing a quick-disconnect solution to your pump line.
- Barb size/tubing int.Ø (port 1):1½"
- Connection Type:Cam to Barb
- Flow pattern:Straight
- Seal material:EPDM
- Description:Cam Fitting, Polypropylene, Female Cam×1½" Hose Barb
- Environmentally Preferable:
- Warranty:1 Month
- Pk:1
- Fitting material:Polypropylene