ContactUsLinkComponent
Masterflex® Adapter Fittings, Luer to Luer, Nylon, Avantor®
Masterflex® Adapter Fittings, Luer to Luer, Nylon, Avantor®
Certificates
undefined
Masterflex® Adapter Fittings, Luer to Luer, Nylon, Avantor®
Certificates
Specifications
  • Model:
  • 45502-84
  • 45502-28
  • 45502-22
  • 45502-85
  • 45502-80
  • 45505-56
  • 45502-56
  • 45505-72
  • Connection type (port 1):
  • Female luer
  • Female luer
  • Female luer
  • Female luer
  • Female luer
  • Male luer lock
  • Female luer
  • Male luer slip
  • Connection type (port 2):
  • Male luer slip
  • Cap
  • Female luer
  • Female luer
  • Male luer lock
  • Plug
  • Female luer
  • Male luer slip
  • Connection type (port 3):
  • Male luer slip
  • Female luer
  • No. of ports:
  • 2
  • 1
  • 2
  • 2
  • 2
  • 1
  • 3
  • 2
  • Warranty:
  • 3 Months
  • 3 Months
  • 3 Months
  • 3 Months
  • 3 Months
  • 3 Months
  • 3 Months
  • 3 Months