ContactUsLinkComponent
Masterflex® Luer Fittings, High-Pressure Stopcock, Avantor®
Masterflex® Fitting, Polycarbonate, Three-Way, High-Pressure Stopcocks with Female Luer (2) to Male Luer Lock, Non-Sterile; 10/PK
Catalog # MFLX30526-34
undefined
Masterflex® Fitting, Polycarbonate, Three-Way, High-Pressure Stopcocks with Female Luer (2) to Male Luer Lock, Non-Sterile; 10/PK
Catalog # MFLX30526-34
Specifications
  • Flow pattern:
    Three-Way
  • Max. pressure:
    1050 psi
  • Description:
    Fitting, Polycarbonate, Three-Way, High-Pressure Stopcocks with Female Luer (2) to Male Luer Lock, Non-Sterile; 10/PK
  • Wetted materials:
    Polycarbonate
  • Warranty:
    Out of box
  • Pk:
    10
  • Inlet port:
    Female Luer(2)
  • Outlet port:
    Male Luer Lock
Specifications
About this item
Pressure rating up to 1050 psi.
  • Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Stopcock fittings feature luer connections.
  • Flow pattern:
    Three-Way
  • Max. pressure:
    1050 psi
  • Description:
    Fitting, Polycarbonate, Three-Way, High-Pressure Stopcocks with Female Luer (2) to Male Luer Lock, Non-Sterile; 10/PK
  • Environmentally Preferable:
  • Wetted materials:
    Polycarbonate
  • Warranty:
    Out of box
  • Pk:
    10
  • Inlet port:
    Female Luer(2)
  • Outlet port:
    Male Luer Lock