ContactUsLinkComponent
CPC® Quick-Disconnect Fittings, Sanitary Bodies and Inserts
CPC® Quick-Disconnect Fittings, Sanitary Bodies and Inserts
Certificates
undefined
CPC® Quick-Disconnect Fittings, Sanitary Bodies and Inserts
Certificates
Specifications
  • Connection type (port 1):
  • Sanitary to coupling insert
  • Sanitary to coupling insert
  • Sanitary to coupling insert
  • Max. temperature:
  • 149 °C (300 °F)
  • 149 °C (300 °F)
  • 149 °C (300 °F)
  • 149 °C (300 °F)
  • 149 °C (300 °F)
  • 149 °C (300 °F)
  • 149 °C (300 °F)
  • 149 °C (300 °F)
  • 149 °C (300 °F)
  • 149 °C (300 °F)
  • 149 °C (300 °F)
  • Min. temperature:
  • –40 °C (–40 °F)
  • –40 °C (–40 °F)
  • –40 °C (–40 °F)
  • –40 °C (–40 °F)
  • –40 °C (–40 °F)
  • –40 °C (–40 °F)
  • –40 °C (–40 °F)
  • –40 °C (–40 °F)
  • –40 °C (–40 °F)
  • –40 °C (–40 °F)
  • –40 °C (–40 °F)
  • O-Ring material:
  • USP Class VI, ADCF
  • USP Class VI Platinum-Cured Silicone
  • USP Class VI Platinum-Cured Silicone
  • USP Class VI Platinum-Cured Silicone
  • USP Class VI Platinum-Cured Silicone
  • USP Class VI Platinum-Cured Silicone
  • USP Class VI, ADCF
  • USP Class VI Platinum-Cured Silicone
  • USP Class VI Platinum-Cured Silicone
  • USP Class VI, ADCF
  • USP Class VI Platinum-Cured Silicone
  • Sterilisation:
  • Autoclave or gamma
  • Autoclave or gamma
  • Autoclave or gamma
  • Autoclave or gamma
  • Autoclave or gamma
  • Autoclave or gamma
  • Autoclave or gamma
  • Autoclave or gamma
  • Autoclave or gamma
  • Autoclave or gamma
  • Autoclave or gamma