ContactUsLinkComponent
Masterflex® Mechanically Coupled Centrifugal Pumps, Avantor®
undefined
Masterflex® Mechanically Coupled Centrifugal Pumps, Avantor®
  • Model:
  • 70725-14
  • 70725-02
  • 70725-06
  • 70725-01
  • 70725-10
  • 70725-07
  • 70725-00
  • Duty cycle continuous:
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Inlet connection:
  • 1,25 NPT(F)
  • 1,25 NPT(F)
  • 1,25 NPT(F)
  • 1,25 NPT(F)
  • 1,25 NPT(F)
  • 1,25 NPT(F)
  • 1,25 NPT(F)
  • Max. pressure:
  • 8,6 bar (125 psi)
  • 8,6 bar (125 psi)
  • 8,6 bar (125 psi)
  • 8,6 bar (125 psi)
  • 8,6 bar (125 psi)
  • 8,6 bar (125 psi)
  • 8,6 bar (125 psi)
  • Max. temperature:
  • 100 °C (212 °F)
  • 100 °C (212 °F)
  • 100 °C (212 °F)
  • 100 °C (212 °F)
  • 100 °C (212 °F)
  • 100 °C (212 °F)
  • 100 °C (212 °F)
  • Max. viscosity:
  • 352 cP
  • 352 cP
  • 352 cP
  • 352 cP
  • 352 cP
  • 352 cP
  • 352 cP
  • Motor type:
  • TEFC
  • TEFC
  • TEFC
  • TEFC
  • TEFC
  • TEFC
  • TEFC
  • Motor hp:
  • 3
  • 1-1/2
  • 2
  • 1-1/2
  • 1-1/2
  • 1
  • 1-1/2
  • Outlet connection:
  • 1 NPT(F)
  • 1 NPT(F)
  • 1 NPT(F)
  • 1 NPT(F)
  • 1 NPT(F)
  • 1 NPT(F)
  • 1 NPT(F)
  • Wetted materials:
  • 304 SS pump head and impeller; SS shaft; Type 21 mechanical seals (carbon vs. ceramic/Viton®); and Viton O-rings
  • 304 SS pump head and impeller; SS shaft; Type 21 mechanical seals (carbon vs. ceramic/Viton®); and Viton O-rings
  • 304 SS pump head and impeller; SS shaft; Type 21 mechanical seals (carbon vs. ceramic/Viton®); and Viton O-rings
  • 304 SS pump head and impeller; SS shaft; Type 21 mechanical seals (carbon vs. ceramic/Viton®); and Viton O-rings
  • 304 SS pump head and impeller; SS shaft; Type 21 mechanical seals (carbon vs. ceramic/Viton®); and Viton O-rings
  • 304 SS pump head and impeller; SS shaft; Type 21 mechanical seals (carbon vs. ceramic/Viton®); and Viton O-rings
  • 304 SS pump head and impeller; SS shaft; Type 21 mechanical seals (carbon vs. ceramic/Viton®); and Viton O-rings
  • Warranty:
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
Frequently Bought Together