Multi-Channel Tubing Racks, Non-Adhesive
An easy solution to secure and route multiple tubing lines.
- Keep your tubing lines organised and untangled
These tube racks offer a simple and economical method for organising tubing lines. The ³/₁₆" and ¹/₄" sizes can be easily separated down to single holders by hand for increased flexibility and custom tubing arrangements. The ³/₁₆" and ¹/₄" sizes are available with adhesive fastener, and all sizes are available with through holes for secure mounting (mounting screws are not included). Standard racks are black polyethylene; ³/₁₆" and ¹/₄" sizes are available in other colours.
- Environmentally Preferable:
- No. of channels:
- 5
- 7
- 10
- 10
- Warranty:
- 1 year
- 1 year
- 1 year
- 1 year
Frequently Bought Together