Orders LinkContactUsLinkComponent
Masterflex® Sanitary Gaskets, Polymers, Avantor®
Masterflex® Sanitary Gaskets, Polymers, Avantor®
Masterflex® Sanitary Gaskets, Polymers, Avantor®
Certificates
Masterflex® Sanitary Gaskets, Polymers, Avantor®
Masterflex® Sanitary Gaskets, Polymers, Avantor®
32 Options Available Below
Product Details & Documents
Ensure a tight seal.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Single-use custom configuration solutions to minimise downtime
These gaskets are for Tri-Clamp® style fittings and come in a wide variety of materials to match specific process needs. All gaskets meet USDA, 3-A and CGMP criteria. FDA Compliant. All silicone, EPDM, PTFE, and Viton® gaskets are animal-derived ingredient free and meet USP Class VI and cytotoxicity criteria.
Platinum-cured silicone is ideal for most pharmaceutical, food, and beverage processing applications. Odorless and tasteless to ensure media purity. May undergo SIP. Usable from –58 to 446 °F (–50 to 230 °C).
Viton offers excellent chemical resistance, especially to many acids and alkalis.
Specifications for Product Family
Specifications
  • Connection type:
  • Sanitary
  • Sanitary
  • Sanitary
  • Sanitary
  • Sanitary
  • Sanitary
  • Sanitary
  • Sanitary
  • Sanitary
  • Sanitary
  • Sanitary
  • Sanitary
  • Sanitary
  • Sanitary
  • Sanitary
  • Sanitary
  • Sanitary
  • Sanitary
  • Sanitary
  • Sanitary
  • Sanitary
  • Sanitary
  • Sanitary
  • Sanitary
  • Sanitary
  • Sanitary
  • Sanitary
  • Sanitary
  • Sanitary
  • Sanitary
  • Sanitary
  • Sanitary
  • Warranty:
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
Specifications
Product Family Options
Product Information
  • Fitting material
    Sanitary clamp size (Port 1)
    Sanitary gasket size
    Availability
    Price
  • MFLX30548-40
    Sanitary Gasket, Buna-N, 3/4" Tri-Clamp®; 10/PK
    Sanitary Gasket, Buna-N, 3/4" Tri-Clamp®; 10/PK
    Buna-N
    3/4"
    3/4"
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 24,90 €
    24,90 €
    1 * 10 ST
       24,90 €
    Specifications:
    More
    Specifications
    Cat. No.
    MFLX30548-40
    Connection type
    Sanitary
    Warranty
    1 year
  • MFLX30548-42
    Sanitary Gasket, Buna-N, 1" Tri-Clamp®; 10/PK
    Sanitary Gasket, Buna-N, 1" Tri-Clamp®; 10/PK
    Buna-N
    1-1/2"
    1"
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 12,30 €
    12,30 €
    1 * 10 ST
       12,30 €
    Specifications:
    More
    Specifications
    Cat. No.
    MFLX30548-42
    Connection type
    Sanitary
    Warranty
    1 year
  • MFLX30548-44
    Sanitary Gasket, Buna-N, 1-1/2" Tri-Clamp®; 10/PK
    Sanitary Gasket, Buna-N, 1-1/2" Tri-Clamp®; 10/PK
    Buna-N
    1-1/2"
    1-1/2"
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 11,80 €
    11,80 €
    1 * 10 ST
       11,80 €
    Specifications:
    More
    Specifications
    Cat. No.
    MFLX30548-44
    Connection type
    Sanitary
    Warranty
    1 year
  • MFLX30548-46
    Sanitary Gasket, Buna-N, 2" Tri-Clamp®; 10/PK
    Sanitary Gasket, Buna-N, 2" Tri-Clamp®; 10/PK
    Buna-N
    2"
    2"
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 10,90 €
    10,90 €
    1 * 10 ST
       10,90 €
    Specifications:
    More
    Specifications
    Cat. No.
    MFLX30548-46
    Connection type
    Sanitary
    Warranty
    1 year
  • MFLX30548-48
    Sanitary Gasket, Buna-N, 2-1/2" Tri-Clamp®; 10/PK
    Sanitary Gasket, Buna-N, 2-1/2" Tri-Clamp®; 10/PK
    Buna-N
    2"
    2-1/2"
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 13,10 €
    13,10 €
    1 * 10 ST
       13,10 €
    Specifications:
    More
    Specifications
    Cat. No.
    MFLX30548-48
    Connection type
    Sanitary
    Warranty
    1 year
  • MFLX29590-80
    Buna N Sanitary Gasket, Black, 4" Tri-Clamp; 1/Pk
    Buna N Sanitary Gasket, Black, 4" Tri-Clamp; 1/Pk
    Buna N
    4"
    4"
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 4,60 €
    4,60 €
    1 * 1 ST
       4,60 €
    Specifications:
    More
    Specifications
    Cat. No.
    MFLX29590-80
    Connection type
    Sanitary
    Warranty
    1 year
  • MFLX30548-60
    Sanitary Gasket, EPDM, 3/4" Tri-Clamp®; 10/PK
    Sanitary Gasket, EPDM, 3/4" Tri-Clamp®; 10/PK
    EPDM
    3/4"
    3/4"
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 25,60 €
    25,60 €
    1 * 10 ST
       25,60 €
    Specifications:
    CE certificate
    More
    Specifications
    Cat. No.
    MFLX30548-60
    Connection type
    Sanitary
    Warranty
    1 year
    CE certificate
  • MFLX30548-62
    Sanitary Gasket, EPDM, 1" Tri-Clamp®; 10/PK
    Sanitary Gasket, EPDM, 1" Tri-Clamp®; 10/PK
    EPDM
    1-1/2"
    1"
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 26,40 €
    26,40 €
    1 * 10 ST
       26,40 €
    Specifications:
    More
    Specifications
    Cat. No.
    MFLX30548-62
    Connection type
    Sanitary
    Warranty
    1 year
  • MFLX30548-64
    Sanitary Gasket, EPDM, 1-1/2" Tri-Clamp®; 10/PK
    Sanitary Gasket, EPDM, 1-1/2" Tri-Clamp®; 10/PK
    EPDM
    1-1/2"
    1-1/2"
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 25,00 €
    25,00 €
    1 * 10 ST
       25,00 €
    Specifications:
    CE certificate
    More
    Specifications
    Cat. No.
    MFLX30548-64
    Connection type
    Sanitary
    Warranty
    1 year
    CE certificate
  • MFLX30548-66
    Sanitary Gasket, EPDM, 2" Tri-Clamp®; 10/PK
    Sanitary Gasket, EPDM, 2" Tri-Clamp®; 10/PK
    EPDM
    2"
    2"
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 26,90 €
    26,90 €
    1 * 10 ST
       26,90 €
    Specifications:
    CE certificate
    More
    Specifications
    Cat. No.
    MFLX30548-66
    Connection type
    Sanitary
    Warranty
    1 year
    CE certificate
  • MFLX30548-68
    Sanitary Gasket, EPDM, 2-1/2" Tri-Clamp®; 10/PK
    Sanitary Gasket, EPDM, 2-1/2" Tri-Clamp®; 10/PK
    EPDM
    2"
    2-1/2"
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 28,60 €
    28,60 €
    1 * 10 ST
       28,60 €
    Specifications:
    CE certificate
    More
    Specifications
    Cat. No.
    MFLX30548-68
    Connection type
    Sanitary
    Warranty
    1 year
    CE certificate
  • MFLX29590-83
    FKM/Viton Sanitary Gasket, 4" Tri-Clamp; 1/Pk
    FKM/Viton Sanitary Gasket, 4" Tri-Clamp; 1/Pk
    FKM/Viton
    4"
    4"
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 19,20 €
    19,20 €
    1 * 1 ST
       19,20 €
    Specifications:
    More
    Specifications
    Cat. No.
    MFLX29590-83
    Connection type
    Sanitary
    Warranty
    1 year
  • MFLX00162-NJ
    Platinum-Cured Silicone Gasket, White, 2"; 10/PK
    Platinum-Cured Silicone Gasket, White, 2"; 10/PK
    Platinum-Cured Silicone
    2"
    2"
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 24,60 €
    24,60 €
    1 * 10 ST
       24,60 €
    Specifications:
    More
    Specifications
    Cat. No.
    MFLX00162-NJ
    Connection type
    Sanitary
    Warranty
    1 year
  • MFLX00364-YQ
    Sanitary Gasket, Platinum-Cured Silicone, 4"; 10/PK
    Sanitary Gasket, Platinum-Cured Silicone, 4"; 10/PK
    Platinum-Cured Silicone
    4"
    4"
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 68,90 €
    68,90 €
    1 * 10 ST
       68,90 €
    Specifications:
    More
    Specifications
    Cat. No.
    MFLX00364-YQ
    Connection type
    Sanitary
    Warranty
    1 year
  • MFLX00404-MO
    Sanitary Gasket, Platinum-Cured Silicone, 6"; 10/PK
    Sanitary Gasket, Platinum-Cured Silicone, 6"; 10/PK
    Platinum-Cured Silicone
    6"
    6"
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 410,00 €
    410,00 €
    1 * 10 ST
       410,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    MFLX00404-MO
    Connection type
    Sanitary
    Warranty
    1 year
  • MFLX30548-00
    Sanitary Gasket, Silicone, 3/4" Tri-Clamp®; 10/PK
    Sanitary Gasket, Silicone, 3/4" Tri-Clamp®; 10/PK
    Platinum-Cured Silicone
    3/4"
    3/4"
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 26,30 €
    26,30 €
    1 * 10 ST
       26,30 €
    Specifications:
    CE certificate
    More
    Specifications
    Cat. No.
    MFLX30548-00
    Connection type
    Sanitary
    Warranty
    1 year
    CE certificate
  • MFLX30548-02
    Sanitary Gasket, Silicone, 1"; 10/PK
    Sanitary Gasket, Silicone, 1"; 10/PK
    Platinum-Cured Silicone
    1-1/2"
    1"
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 36,20 €
    36,20 €
    1 * 10 ST
       36,20 €
    Specifications:
    CE certificate
    More
    Specifications
    Cat. No.
    MFLX30548-02
    Connection type
    Sanitary
    Warranty
    1 year
    CE certificate
  • MFLX30548-04
    Sanitary Gasket, Silicone, 1-1/2"; 10/PK
    Sanitary Gasket, Silicone, 1-1/2"; 10/PK
    Platinum-Cured Silicone
    1-1/2"
    1-1/2"
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 26,30 €
    26,30 €
    1 * 10 ST
       26,30 €
    Specifications:
    CE certificate
    More
    Specifications
    Cat. No.
    MFLX30548-04
    Connection type
    Sanitary
    Warranty
    1 year
    CE certificate
  • MFLX30548-06
    Sanitary Gasket, Silicone, 2"; 10/PK
    Sanitary Gasket, Silicone, 2"; 10/PK
    Platinum-Cured Silicone
    2"
    2"
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 36,20 €
    36,20 €
    1 * 10 ST
       36,20 €
    Specifications:
    CE certificate
    More
    Specifications
    Cat. No.
    MFLX30548-06
    Connection type
    Sanitary
    Warranty
    1 year
    CE certificate
  • MFLX30548-08
    Sanitary Gasket, Silicone, 2-1/2"; 10/PK
    Sanitary Gasket, Silicone, 2-1/2"; 10/PK
    Platinum-Cured Silicone
    2"
    2-1/2"
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 40,00 €
    40,00 €
    1 * 10 ST
       40,00 €
    Specifications:
    CE certificate
    More
    Specifications
    Cat. No.
    MFLX30548-08
    Connection type
    Sanitary
    Warranty
    1 year
    CE certificate
  • MFLX41001-77
    Sanitary Gasket, Platinum-Cured Silicone, 3"; 10/PK
    Sanitary Gasket, Platinum-Cured Silicone, 3"; 10/PK
    Platinum-Cured Silicone
    3"
    3"
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 46,10 €
    46,10 €
    1 * 10 ST
       46,10 €
    Specifications:
    More
    Specifications
    Cat. No.
    MFLX41001-77
    Connection type
    Sanitary
    Warranty
    1 year
  • MFLX30548-80
    Sanitary Gasket, PTFE, 3/4" Tri-Clamp®; 10/PK
    Sanitary Gasket, PTFE, 3/4" Tri-Clamp®; 10/PK
    PTFE
    3/4"
    3/4"
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 31,40 €
    31,40 €
    1 * 10 ST
       31,40 €
    Specifications:
    CE certificate
    More
    Specifications
    Cat. No.
    MFLX30548-80
    Connection type
    Sanitary
    Warranty
    1 year
    CE certificate
  • MFLX30548-82
    Sanitary Gasket, PTFE, 1" Tri-Clamp®; 10/PK
    Sanitary Gasket, PTFE, 1" Tri-Clamp®; 10/PK
    PTFE
    1-1/2"
    1"
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 41,40 €
    41,40 €
    1 * 10 ST
       41,40 €
    Specifications:
    CE certificate
    More
    Specifications
    Cat. No.
    MFLX30548-82
    Connection type
    Sanitary
    Warranty
    1 year
    CE certificate
  • MFLX30548-84
    Sanitary Gasket, PTFE, 1-1/2" Tri-Clamp®; 10/PK
    Sanitary Gasket, PTFE, 1-1/2" Tri-Clamp®; 10/PK
    PTFE
    1-1/2"
    1-1/2"
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 38,30 €
    38,30 €
    1 * 10 ST
       38,30 €
    Specifications:
    CE certificate
    More
    Specifications
    Cat. No.
    MFLX30548-84
    Connection type
    Sanitary
    Warranty
    1 year
    CE certificate
  • MFLX30548-86
    Sanitary Gasket, PTFE, 2" Tri-Clamp®; 10/PK
    Sanitary Gasket, PTFE, 2" Tri-Clamp®; 10/PK
    PTFE
    2"
    2"
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 40,00 €
    40,00 €
    1 * 10 ST
       40,00 €
    Specifications:
    CE certificate
    More
    Specifications
    Cat. No.
    MFLX30548-86
    Connection type
    Sanitary
    Warranty
    1 year
    CE certificate
  • MFLX30548-88
    Sanitary Gasket, PTFE, 2-1/2" Tri-Clamp®; 10/PK
    Sanitary Gasket, PTFE, 2-1/2" Tri-Clamp®; 10/PK
    PTFE
    2"
    2-1/2"
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 43,90 €
    43,90 €
    1 * 10 ST
       43,90 €
    Specifications:
    CE certificate
    More
    Specifications
    Cat. No.
    MFLX30548-88
    Connection type
    Sanitary
    Warranty
    1 year
    CE certificate
  • MFLX16011-66
    Silicone Sanitary Gasket, 0,5" Tri-Clamp; 10/Pk
    Silicone Sanitary Gasket, 0,5" Tri-Clamp; 10/Pk
    Silicone
    3/4"
    0,5"
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 27,50 €
    27,50 €
    1 * 10 ST
       27,50 €
    Specifications:
    More
    Specifications
    Cat. No.
    MFLX16011-66
    Connection type
    Sanitary
    Warranty
    1 year
  • MFLX30548-20
    Sanitary Gasket, Viton®, 3/4" Tri-Clamp®; 10/PK
    Sanitary Gasket, Viton®, 3/4" Tri-Clamp®; 10/PK
    Viton®
    3/4"
    3/4"
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 51,70 €
    51,70 €
    1 * 10 ST
       51,70 €
    Specifications:
    CE certificate
    More
    Specifications
    Cat. No.
    MFLX30548-20
    Connection type
    Sanitary
    Warranty
    1 year
    CE certificate
  • MFLX30548-24
    Sanitary Gasket, Viton®, 1-1/2" Tri-Clamp®; 10/PK
    Sanitary Gasket, Viton®, 1-1/2" Tri-Clamp®; 10/PK
    Viton®
    1-1/2"
    1-1/2"
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 51,00 €
    51,00 €
    1 * 10 ST
       51,00 €
    Specifications:
    CE certificate
    More
    Specifications
    Cat. No.
    MFLX30548-24
    Connection type
    Sanitary
    Warranty
    1 year
    CE certificate
  • MFLX30548-26
    Sanitary Gasket, Viton®, 2" Tri-Clamp®; 10/PK
    Sanitary Gasket, Viton®, 2" Tri-Clamp®; 10/PK
    Viton®
    2"
    2"
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 52,60 €
    52,60 €
    1 * 10 ST
       52,60 €
    Specifications:
    CE certificate
    More
    Specifications
    Cat. No.
    MFLX30548-26
    Connection type
    Sanitary
    Warranty
    1 year
    CE certificate
  • MFLX30548-28
    Sanitary Gasket, Viton®, 2-1/2" Tri-Clamp®; 10/PK
    Sanitary Gasket, Viton®, 2-1/2" Tri-Clamp®; 10/PK
    Viton®
    2"
    2-1/2"
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 61,10 €
    61,10 €
    1 * 10 ST
       61,10 €
    Specifications:
    CE certificate
    More
    Specifications
    Cat. No.
    MFLX30548-28
    Connection type
    Sanitary
    Warranty
    1 year
    CE certificate
  • MFLX30548-22
    Sanitary Gasket, Viton®, 1" Tri-Clamp®; 10/PK
    Sanitary Gasket, Viton®, 1" Tri-Clamp®; 10/PK
    Viton®
    1-1/2"
    1"
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 56,80 €
    56,80 €
    1 * 10 ST
       56,80 €
    Specifications:
    CE certificate
    More
    Specifications
    Cat. No.
    MFLX30548-22
    Connection type
    Sanitary
    Warranty
    1 year
    CE certificate