ContactUsLinkComponent
Masterflex® L/S® Standard Long-Shaft Pump Heads, Avantor®
Masterflex® L/S® Standard Long-Shaft Pump Heads, Avantor®
Certificates
undefined
Masterflex® L/S® Standard Long-Shaft Pump Heads, Avantor®
Specifications
  • Model:
  • 07024-10
  • 07024-00
  • 07015-50
  • 07016-10
  • 07015-10
  • 07013-10
  • 07016-00
  • 07015-00
  • Gear ratio:
  • 1:1
  • 1:1
  • 1:1
  • 1:1
  • 1:1
  • 1:1
  • Min. flow rate:
  • 2,8 ml/min
  • 2,8 ml/min
  • 1,7 ml/min
  • 0,8 ml/min
  • 1,7 ml/min
  • 0,06 ml/min
  • 0,8 ml/min
  • 1,7 ml/min
  • No. of channels:
  • 1
  • 1
  • 1
  • 1
  • 1
  • 1
  • Occlusion:
  • Fixed
  • Fixed
  • Fixed
  • Fixed
  • Fixed
  • Fixed
  • Optimum RPM range:
  • 0,02 - 600 rpm
  • 0,02 - 600 rpm
  • 0,02 - 600 rpm
  • 0,02 - 600 rpm
  • 0,02 - 600 rpm
  • 0,02 - 600 rpm
  • Pump series:
  • L/S
  • L/S
  • L/S
  • L/S
  • L/S
  • L/S
  • L/S
  • L/S
  • Tubing retention:
  • Manual
  • Manual
  • Manual
  • Manual
  • Manual
  • Manual
  • Tubing sizes accepted:
  • L/S® 24
  • L/S® 24
  • L/S® 15
  • L/S® 16
  • L/S® 15
  • L/S® 13
  • L/S® 16
  • L/S® 15
  • Volume per revolution with L/S 13 tubing:
  • 0,06 ml
  • Volume per revolution with L/S 15 tubing:
  • 1,7 ml
  • 1,7 ml
  • 1,7 ml
  • Volume per revolution with L/S 16 tubing:
  • 0,8 ml
  • 0,8 ml
  • Volume per revolution with L/S 24 tubing:
  • 2,8 ml
  • 2,8 ml
  • Warranty:
  • 2 years
  • 2 years
  • 2 years
  • 2 years
  • 2 years
  • 2 years
  • 2 years
  • 2 years