ContactUsLinkComponent
Masterflex® L/S® Easy-Load® 3 Pump Heads for High-Performance Precision Tubing, Avantor®
Masterflex® L/S® Easy-Load® 3 Pump Heads for High-Performance Precision Tubing, Avantor®
Certificates
undefined
Masterflex® L/S® Easy-Load® 3 Pump Heads for High-Performance Precision Tubing, Avantor®
Specifications
  • Gear ratio:
  • 1:1
  • 1:1
  • No. of channels:
  • 1
  • 1
  • Occlusion:
  • Fixed
  • Fixed
  • Open-head sensor:
  • No
  • No
  • Optimum RPM range:
  • 0,02 - 600 rpm
  • 0,02 - 600 rpm
  • Pump series:
  • L/S
  • L/S
  • Suction lift:
  • 8,8 m (29 ft.)
  • 8,8 m (29 ft.)
  • Tubing retention:
  • Automatic
  • Automatic
  • Tubing sizes accepted:
  • L/S® 15, L/S 24, L/S 35, L/S 36
  • L/S® 15, L/S 24, L/S 35, L/S 36
  • Volume per revolution with L/S 15 tubing:
  • 1,7 ml
  • 1,7 ml
  • Volume per revolution with L/S 24 tubing:
  • 2,8 ml
  • 2,8 ml
  • Volume per revolution with L/S 35 tubing:
  • 3,8 ml
  • 3,8 ml
  • Volume per revolution with L/S 36 tubing:
  • 4,8 ml
  • 4,8 ml
  • Warranty:
  • 2 years
  • 2 years