ContactUsLinkComponent
Battery-Powered Digital Flow Rate Monitor and Totalizer 43643
GF Signet Digital Flow Rate Monitor/Totalizer; Battery Powered
Catalog # MFLX05629-22
CE Compliant
CSA Certified
IP65 washdown
UL Listed
ISO9001
undefined
GF Signet Digital Flow Rate Monitor/Totalizer; Battery Powered
Catalog # MFLX05629-22
CE Compliant
CSA Certified
IP65 washdown
UL Listed
ISO9001
Specifications
  • Accuracy:
    ±0,5% of reading
  • Display type:
    4-digit LCD (upper). Lower: 8-digit LCD
  • Description:
    3-8150-1P Digital Flow Rate Monitor/ Totalizer Battery Powered
  • Battery life:
    4 years nominal
  • Max. operating temperature:
    55 °C (131 °F)
  • Warranty:
    2 years
  • Pk:
    1
Specifications
About this item
Use in areas where AC power is not readily available.
  • Use in areas where wired power is power is not readily available
Independently calibrate flow and totalizer readings. Monitor includes both fixed and resettable totalizers. Features a four-digit flow LCD, and eight-digit totalizer LCD, automatic contrast adjustment, and low-battery indication. Monitor includes unit/multiplier decals for field calibration.
Operates on two internal lithium batteries (included). Use monitor only with the Roto-X™ and Metalex™ flow sensors sold separately. Note: not for use with the Roto-X™ Low-Flow sensors.
  • Accuracy:
    ±0,5% of reading
  • Display type:
    4-digit LCD (upper). Lower: 8-digit LCD
  • Description:
    3-8150-1P Digital Flow Rate Monitor/ Totalizer Battery Powered
  • Environmentally Preferable:
  • Battery life:
    4 years nominal
  • Max. operating temperature:
    55 °C (131 °F)
  • Warranty:
    2 years
  • Pk:
    1