ContactUsLinkComponent
Masterflex® Liquid Flow Switches for High Inline Pressures, Avantor®
Masterflex® Liquid Flow Switches for High Inline Pressures, Avantor®
Certificates
undefined
Masterflex® Liquid Flow Switches for High Inline Pressures, Avantor®
Specifications
  • Model:
  • 32774-40
  • 32774-32
  • 32774-48
  • Max. pressure:
  • 1500 psi
  • 1500 psi
  • 1500 psi
  • Max. operating temperature:
  • 135 °C (275 °F)
  • 135 °C (275 °F)
  • 135 °C (275 °F)
  • Wetted materials:
  • Stainless Steel, PPS composite, 316 SS, and fluorocarbon
  • Brass, PPS composite, 316 SS, and fluorocarbon
  • Stainless Steel, PPS composite, 316 SS, and fluorocarbon
  • Process connection:
  • 0,375 NPT(M)
  • 0,375 NPT(M)
  • 0,375 NPT(M)
  • Height:
  • 40751"
  • 40751"
  • 40751"
  • Warranty:
  • 1 year
  • 1 year
  • 1 year