Masterflex® L/S® High-Performance Pump Head Tubing Retainer Kit
MFLX77250-66
- Pump series:L/S
- Description:Tubing retainer kit for L/S high-performance pump head
- Warranty:2 years
- Pk:1
Extend the life of your Masterflex pump head with this service kit.
- Tubing retainer prevents tubing from moving within the pump head
Compatible with the Masterflex® L/S® High-Performance Pump Heads 77250-62.
- Pump series:L/S
- Description:Tubing retainer kit for L/S high-performance pump head
- Environmentally Preferable:
- Warranty:2 years
- Pk:1