ContactUsLinkComponent
Masterflex® I/P® High-Performance Pump Head Replacement Parts, Avantor®
Masterflex® I/P® High-Performance Pump Head Replacement Rotor and Body Assembly
Catalog # MFLX77600-69
undefined
Masterflex® I/P® High-Performance Pump Head Replacement Rotor and Body Assembly
Catalog # MFLX77600-69
Specifications
  • Pump series:
    I/P
  • Description:
    KIT REPL ROTOR & BODY ASSEMBLY (Rotor/body assembly with tang boot)
  • Pk:
    1
  • Housing material:
    Stainless steel, thermoset polyester
  • Rotor material:
    Stainless Steel
Specifications
About this item
Get the most our of your Masterflex® I/P® High-Performance Pump Head with these replacement parts.
  • Stainless steel (SS) rotor / roller for greater durability and chemical resistance
Replacement parts to keep your pump head operating even longer.
Delivery: Includes rotor, body, and tang boot.
  • Pump series:
    I/P
  • Description:
    KIT REPL ROTOR & BODY ASSEMBLY (Rotor/body assembly with tang boot)
  • Environmentally Preferable:
  • Pk:
    1
  • Housing material:
    Stainless steel, thermoset polyester
  • Rotor material:
    Stainless Steel