ContactUsLinkComponent
AW-Lake Gear Flow Sensor Flow Monitor Transmitters
AW-Lake Gear Flow Sensor/Transmitter Accessory, Flow Monitor w/ 4-20 mA Output
Catalog # MFLX32928-76
undefined
AW-Lake Gear Flow Sensor/Transmitter Accessory, Flow Monitor w/ 4-20 mA Output
Catalog # MFLX32928-76
Specifications
  • Output:
    4 to 20 mA
  • Power:
    110 VAC
  • Display type:
    Digital
  • Description:
    Gear Flow Sensor/Transmitter Accessory, Flow Monitor w/ 4-20 mA Output
  • Warranty:
    1 year
  • Pk:
    1
Specifications
About this item
Easily control flow of your gear flow sensors.
  • Monitor both flow rate and total flow
Monitor both flow rate and flow from pulse output flowmeters in any engineering unit with a push of a button. Programmable, compact, and easy-to-use. Input square wave, sine, or sawtooth frequency. Back-up totalizer lock-out feature prevents accidental erasure. High-speed microprocessor ensures fast and accurate reporting — includes a 4 to 20 mA output for recording or datalogging. Model 32928-76 has two channels to monitor dual flows and display them separately, as a sum, as a difference, or as a ratio.
  • Output:
    4 to 20 mA
  • Power:
    110 VAC
  • Display type:
    Digital
  • Description:
    Gear Flow Sensor/Transmitter Accessory, Flow Monitor w/ 4-20 mA Output
  • Environmentally Preferable:
  • Warranty:
    1 year
  • Pk:
    1