Order Entry
Austria
ContactUsLinkComponent
Hand-Operated FDA Compliant Drum Pumps
Hand-operated FDA-compliant drum pump with outlet hose
  MFLX70607-20
undefined
Hand-operated FDA-compliant drum pump with outlet hose
  MFLX70607-20

 

  • Description:
    Hand-operated FDA-compliant drum pump with outlet hose
  • Wetted materials:
    PVC barrel and hose, Viton® valves and seals, LDPE tube
  • Inlet connection:
    3/4" NPT(M)
  • Tube ext.Ø, outlet:
    1,25"
  • Tube length, inlet:
    12"
  • Tube length, outlet:
    12"
  • Pk:
    1

 

 

Ideal for most acid/base or ionic salt applications.

  • Materials are FDA compliant for indirect food contact
  • Available with outlet hose (70607-20) or spout (70607-22)

These plastic hand-operated drum pumps are constructed with materials that conform under FDA CFR 21 regulations for indirect food contact. Pumps features 26 strokes/gallon (7 strokes/liter) with 3/4” NPT(M) container adapter. Both pumps are constructed with PVC barrel, Viton® valves and seals, and LDPE tube. Choose between models with a PVC hose outlet or LDPE spout outlet.

  • Description:
    Hand-operated FDA-compliant drum pump with outlet hose
  • Environmentally Preferable:
  • Wetted materials:
    PVC barrel and hose, Viton® valves and seals, LDPE tube
  • Inlet connection:
    3/4" NPT(M)
  • Tube ext.Ø, outlet:
    1,25"
  • Tube length, inlet:
    12"
  • Tube length, outlet:
    12"
  • Pk:
    1