ContactUsLinkComponent
Masterflex® Electro-Optic Point-Level Sensors, Avantor®
Masterflex® Electro-Optic Point-Level Sensors, Avantor®
Certificates
undefined
Masterflex® Electro-Optic Point-Level Sensors, Avantor®
Specifications
  • Model:
  • 32772-30
  • 32772-22
  • 32772-32
  • 32772-10
  • 32772-20
  • 32772-12
  • 32772-02
  • 32772-00
  • 32772-04
  • 32772-08
  • Max. process temperature:
  • 100 °C
  • 80 °C
  • 100 °C
  • 80 °C
  • 80 °C
  • 100 °C
  • 80 °C
  • 80 °C
  • 80 °C
  • 80 °C
  • Max. pressure:
  • 2500 psi
  • 150 psi
  • 2500 psi
  • 150 psi
  • 150 psi
  • 150 psi
  • 150 psi
  • 150 psi
  • 150 psi
  • 150 psi
  • Process mount:
  • 1/2"NPT(M)
  • Flanged
  • 1/2"NPT(M)
  • 1/2" Straight Thread
  • Flanged
  • 3/8" NPT(M)
  • 1/4" NPT(M)
  • 1/4" NPT(M)
  • 1/4" NPT(M)
  • 1/2" Straight Thread
  • Relay output:
  • 30 VDC at 40 mA max
  • 30 VDC at 40 mA max
  • 30 VDC at 40 mA max
  • 30 VDC at 40 mA max
  • 30 VDC at 40 mA max
  • 30 VDC at 40 mA max
  • 30 VDC at 40 mA max
  • 30 VDC at 40 mA max
  • 30 VDC at 40 mA max
  • 30 VDC at 40 mA max
  • Wetted materials:
  • Nickel-plated Carbon steel/Fused glass
  • Polysulfone
  • Nickel-plated Carbon steel/Fused glass
  • Polysulfone
  • Polysulfone
  • Isoplast
  • Polysulfone
  • Polysulfone
  • Nylon
  • Polysulfone
  • Warranty:
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year