Order Entry
Austria
ContactUsLinkComponent
Microcentrifuge tube rack, Axygen®
Microcentrifuge tube rack, Axygen®
Catalog # AXYGTR8210
Supplier:  Corning
undefined
Microcentrifuge tube rack, Axygen®
Catalog # AXYGTR8210
Supplier:  Corning

Specifications

  • For tubes:
    20×1,5/2,0 ml
  • Description:
    Microcentrifuge tube rack, 75×210×35 mm (2,9×8,3×1,4")
  • Pk:
    2

Specifications

About this item

Microcentrifuge tube rack is molded from durable, autoclavable polypropylene, and features non-skid rubber feet.

  • Step design allows easy access to tubes
  • Two rows of 10 (20×1,5 to 2,0 ml total)

The rack has two levels and exhibits wide spacing to provide prime sample access and visibility. Tube positions are numbered 1 to 20 for organizing samples.

Packaging: Two in a case.