Order Entry
Austria
ContactUsLinkComponent
AxyPrep™ FragmentSelect-I kit, Axygen®
AxyPrep™ FragmentSelect-I kit, Axygen®
Supplier:  Corning
Certificates
undefined
AxyPrep™ FragmentSelect-I kit, Axygen®
Supplier:  Corning
Certificates

Specifications

  • Method / Format:
    Magnetic beads
  • Prep size:
    Scalable
  • Refine target molecule:
    Amplified/modified DNA
  • Sample type:
    Sequencing reactions/libraries
  • Target molecule:
    DNA

Specifications

Related Information

Frequently Bought Together