Order Entry
Austria
ContactUsLinkComponent
Phosphate-buffered saline (PBS), Corning®
Phosphate-buffered saline (PBS), Corning®
Supplier:  Corning
Certificates
undefined
Phosphate-buffered saline (PBS), Corning®
Supplier:  Corning
Certificates

Specifications

  • Mycoplasma:
    Tested Negative
  • Endotoxin content:
    < 0.25 EU/mL
  • Shelf life:
    24 months
  • Storage temperature:
    2...30 °C

Specifications

Frequently Bought Together