ContactUsLinkComponent
384-well microplate black/clear, round bottom, ultra-low attachment, spheroid, Corning®
384-well microplate black/clear, round bottom, ultra-low attachment, spheroid, Corning®
Catalog # CORN3830BC
Supplier:  Corning
   Low Manufacturing Impact
undefined
384-well microplate black/clear, round bottom, ultra-low attachment, spheroid, Corning®
Catalog # CORN3830BC
Supplier:  Corning
   Low Manufacturing Impact
Specifications
  • No. of wells:
    384
  • Packed:
    10/pack, 50/case
  • Recommended working volume:
    20 - 80 µl
  • Sterile:
    +
  • Volume:
    90 µl
  • Colour:
    Black/clear
  • Description:
    Round bottom, 384 well
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Environmental certification:
    RECs, ISO 14001 - Kennebunk
  • Culture area:
    0,06 cm²
  • Pk:
    50
Specifications
About this item
Ultra-Low Attachment microplates feature a covalently bound hydrogel layer that effectively inhibits cellular attachment.
  • Sterilised by gamma irradiation
  • Opaque walls to prevent well-to-well crosstalk
  • Surface is noncytotoxic, biologically inert and nondegradable
  • With Generic Bar codes, and lid
Surface minimises protein absorption, enzyme activation and cellular activation; Black wall microplates have low background fluorescence, minimal light scatter and reduced crosstalk.
Can be used for both top and bottom reading instruments. Novel well geometry aids formation of spheroids in center of well. Optically clear round bottom with black opaque microplate body
Surface treatment: Ultra-low attachment.
Packaging: Bulk packed 10 per bag.