Order Entry
Austria
ContactUsLinkComponent
Mag PCR normalizer kit, Axygen® AxyPrep
Mag PCR normalizer kit, Axygen® AxyPrep
Supplier:  Corning
Certificates
undefined
Mag PCR normalizer kit, Axygen® AxyPrep
Supplier:  Corning
Certificates

Specifications

  • Method / Format:
    Magnetic beads
  • Prep size:
    Scalable
  • Refine target molecule:
    Amplified/modified DNA
  • Sample type:
    PCR and other enzymatic reactions
  • Target molecule:
    DNA
  • Description:
    AxyPrep™ Mag PCR normalizer kit

Specifications

Related Information

Frequently Bought Together