Order Entry
Austria
ContactUsLinkComponent
Mag plasmid kit, Axygen® AxyPrep™
Mag plasmid kit, Axygen® AxyPrep™
Catalog # AXYGMAG-P-10
Supplier:  Corning
CAS Number:  
Mag plasmid kit, Axygen® AxyPrep™
Catalog # AXYGMAG-P-10
Supplier:  Corning
CAS Number:  

Specifications

  • Pk:
    1
  • Method / Format:
    Magnetic beads
  • Prep size:
    Scalable
  • Refine target molecule:
    Plasmid
  • Sample type:
    Bacteria
  • Target molecule:
    Plasmid
  • Description:
    AxyPrep™ Mag plasmid kit, 10 preps

Specifications

About this item

The AxyPrep™ Mag Plasmid purification kit utilises a magnetic bead technology for high-throughput purification of plasmid DNA from E.coli cells.

AxyPrep™ Mag Plasmid kit can also be used with fosmid and BAC vector-based constructs ranging from 5 to 150kb.