ContactUsLinkComponent
ITT Jabsco Positive Displacement Pump Pulse Dampeners
ITT Jabsco Polypropylene Pulse Dampeners With Santoprene Diaphragm, 3/8" NPT
Catalog # MFLX74021-04
undefined
ITT Jabsco Polypropylene Pulse Dampeners With Santoprene Diaphragm, 3/8" NPT
Catalog # MFLX74021-04
Specifications
  • Description:
    Polypropylene Pulse Dampeners With Santoprene Diaphragm, 3/8" NPT
  • Pk:
    1
  • Wetted materials:
    Polypropylene, Santoprene®
  • Air inlet connection:
    0,25 NPT(F)
  • Inlet connection:
    0,375 NPT(F)
  • Outlet connection:
    0,375 NPT(F)
  • Warranty:
    1 year
Specifications
About this item
Create a pulse-free flow out of your positive displacement pump.
  • Choose between PTFE or Polypropylene with Santoprene dampners
Dampeners work by pre-filling the chamber with compressed air or other gas, then as fluid pressure increases it compresses the air/gas in the chamber. When the air/gas pressure exceeds the fluid pressure the fluid is forced back out of the dampener in between pulse cycles, resulting in a near pulseless flow.
More Info: These pulse dampeners can be used with any positive displacement pump that needs flow with reduced pulsation.
  • Description:
    Polypropylene Pulse Dampeners With Santoprene Diaphragm, 3/8" NPT
  • Environmentally Preferable:
  • Pk:
    1
  • Wetted materials:
    Polypropylene, Santoprene®
  • Air inlet connection:
    0,25 NPT(F)
  • Inlet connection:
    0,375 NPT(F)
  • Outlet connection:
    0,375 NPT(F)
  • Warranty:
    1 year