ContactUsLinkComponent
CPC® ChemQuik® Quick-Disconnect Fittings, In-Line Coupling Bodies
CPC® ChemQuik® Quick-Disconnect Fittings, In-Line Coupling Bodies
Certificates
undefined
CPC® ChemQuik® Quick-Disconnect Fittings, In-Line Coupling Bodies
Specifications
  • Max. operating pressure:
  • 80 psi
  • 80 psi
  • 80 psi
  • Max. temperature:
  • 107 °C (225 °F)
  • 107 °C (225 °F)
  • 107 °C (225 °F)
  • Min. temperature:
  • 0 °C (32 °F)
  • 0 °C (32 °F)
  • 0 °C (32 °F)
  • Seal material:
  • Viton®
  • Viton®
  • Viton®
  • Valve spring material:
  • PEEK
  • PEEK
  • PEEK
  • Warranty:
  • 3 Months
  • 3 Months
  • 3 Months