ContactUsLinkComponent
Masterflex® Magnetic Drive Centrifugal Pump, Avantor®
Masterflex® Magnetic Drive Centrifugal Pump, Avantor®
Certificates
undefined
Masterflex® Magnetic Drive Centrifugal Pump, Avantor®
Specifications
  • Model:
  • 75226-10
  • 75226-35
  • 75226-32
  • 75226-15
  • 75226-12
  • Duty cycle continuous:
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Inlet connection:
  • 1 NPT(F)
  • 1,5 NPT(F)
  • 1,5 NPT(F)
  • 1 NPT(F)
  • 1 NPT(F)
  • Max. pressure:
  • 7,6 bar (110 psi)
  • 7,6 bar (110 psi)
  • 7,6 bar (110 psi)
  • 7,6 bar (110 psi)
  • 7,6 bar (110 psi)
  • Max. temperature:
  • 71 °C (160 °F)
  • 71 °C (160 °F)
  • 71 °C (160 °F)
  • 71 °C (160 °F)
  • 71 °C (160 °F)
  • Max. viscosity:
  • 140 cP
  • 140 cP
  • 140 cP
  • 140 cP
  • 140 cP
  • Motor type:
  • TEFC
  • TEFC
  • TEFC
  • TEFC
  • Motor hp:
  • 1
  • 1
  • 1/2
  • 1/2
  • Wetted materials:
  • PP pump head and impeller, ceramic shaft, carbon fiber-filled PTFE thrust ring and sleeve bearings; EPDM (Nordel) O-ring
  • PP pump head and impeller, ceramic shaft, carbon fiber-filled PTFE thrust ring and sleeve bearings; EPDM (Nordel) O-ring
  • PP pump head and impeller, ceramic shaft, carbon fiber-filled PTFE thrust ring and sleeve bearings; EPDM (Nordel) O-ring
  • PP pump head and impeller, ceramic shaft, carbon fiber-filled PTFE thrust ring and sleeve bearings; EPDM (Nordel) O-ring
  • PP pump head and impeller, ceramic shaft, carbon fiber-filled PTFE thrust ring and sleeve bearings; EPDM (Nordel) O-ring
  • Warranty:
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year