ContactUsLinkComponent
John Guest® Speedfit Ball Valves, Polypropylene
John Guest® Speedfit Ball Valves, Polypropylene
Certificates
undefined
John Guest® Speedfit Ball Valves, Polypropylene
Specifications
  • Max. pressure:
  • 10,3 bar (150 psi)
  • 10,3 bar (150 psi)
  • 10,3 bar (150 psi)
  • 10,3 bar (150 psi)
  • 10,3 bar (150 psi)
  • 10,3 bar (150 psi)
  • Max. temperature:
  • 65 °C (150 °F)
  • 65 °C (150 °F)
  • 65 °C (150 °F)
  • 65 °C (150 °F)
  • 65 °C (150 °F)
  • 65 °C (150 °F)
  • Wetted materials:
  • food-grade polypropylene, EPDM, and stainless steel
  • food-grade polypropylene, EPDM, and stainless steel
  • food-grade polypropylene, EPDM, and stainless steel
  • food-grade polypropylene, EPDM, and stainless steel
  • food-grade polypropylene, EPDM, and stainless steel
  • food-grade polypropylene, EPDM, and stainless steel
  • Warranty:
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year