ContactUsLinkComponent
Masterflex® PTFE Ball Valves, Female NPT, Avantor®
Masterflex® PTFE Ball Valves, Female NPT, Avantor®
Certificates
undefined
Masterflex® PTFE Ball Valves, Female NPT, Avantor®
Specifications
  • Model:
  • 98514-09
  • 98514-05
  • 98514-01
  • Max. pressure:
  • 4,1 bar (60 psi)
  • 4,1 bar (60 psi)
  • 5,5 bar (80 psi)
  • Max. temperature:
  • 70 °C (158 °F)
  • 70 °C (158 °F)
  • 70 °C (158 °F)
  • Wetted materials:
  • PTFE
  • PTFE
  • PTFE
  • Warranty:
  • 1 year
  • 1 year
  • 1 year