Order Entry
Austria
ContactUsLinkComponent
 
Masterflex® I/P® Variable-Speed Air-Powered Drive with Easy-Load® ATEX Pump Head, 100 to 650 rpm
  MFLX77980-10
CE Compliant
undefined
Masterflex® I/P® Variable-Speed Air-Powered Drive with Easy-Load® ATEX Pump Head, 100 to 650 rpm
  MFLX77980-10
CE Compliant

 

  • Maximum Flow Rate:
    8000 ml/min
  • Accuracy:
    ±10%
  • Control type:
    Variable-Speed Air-Powered
  • Min. flow rate:
    1200 ml/min
  • No. of channels:
    1
  • Pump series:
    I/P
  • Description:
    I/P ATEX Zone 2 Air-Powered Pump System, 100-650 rpm.
  • Height:
    22,86 cm
  • Length:
    33,35 cm
  • Seq:
    02
  • Model:
    77980-10
  • Duty cycle:
    Continuous
  • Drive IP rating:
    IP 34
  • Min. RPM:
    100 rpm
  • Open-head sensor:
    No
  • Tubing sizes accepted:
    I/P® 26, 73, 82
  • Washdown capable:
    No
  • Tubing:
    06434-73, 10-ft (C-FLEX® Ultra, I/P® 73)
  • Drive:
    07589-30 (I/P® Variable-Speed Air-Powered Drive)
  • No. of rollers:
    3
  • Warranty:
    2 years
  • Pk:
    1
  • Pump head:
    77602-80 (I/P® Easy-Load® ATEX Head)
  • Width:
    21,59 cm
  • Housing material:
    Painted steel
  • Max. RPM:
    650 rpm

 

 

Pump safely in hazardous
environments.

  • Flow rate 0.6 to 13 LPM I/P tubing—flow rate depends on drive rpm, pump head, and tubing size
  • Easily adjust motor speed by using included regulator
  • Effortlessly connect to your existing air supply or compressor
  • The 3/4-hp air-powered motor starts smooth, runs cool, and requires little maintenance
  • ATEX Zone 2 approved; rated to EEXII 3 G c IIC T6; NEC Class 1, Division 2, Groups A, B, C, D, T6

Use this variable-speed air-powered drive when electrical power may be unsafe or impractical. For safe operation, carefully ground to protect from static electricity.

  • Maximum Flow Rate:
    8000 ml/min
  • Accuracy:
    ±10%
  • Control type:
    Variable-Speed Air-Powered
  • Min. flow rate:
    1200 ml/min
  • No. of channels:
    1
  • Pump series:
    I/P
  • Description:
    I/P ATEX Zone 2 Air-Powered Pump System, 100-650 rpm.
  • Height:
    22,86 cm
  • Length:
    33,35 cm
  • Seq:
    02
  • Environmentally Preferable:
  • Model:
    77980-10
  • Duty cycle:
    Continuous
  • Drive IP rating:
    IP 34
  • Min. RPM:
    100 rpm
  • Open-head sensor:
    No
  • Tubing sizes accepted:
    I/P® 26, 73, 82
  • Washdown capable:
    No
  • Tubing:
    06434-73, 10-ft (C-FLEX® Ultra, I/P® 73)
  • Drive:
    07589-30 (I/P® Variable-Speed Air-Powered Drive)
  • No. of rollers:
    3
  • Warranty:
    2 years
  • Pk:
    1
  • Pump head:
    77602-80 (I/P® Easy-Load® ATEX Head)
  • Width:
    21,59 cm
  • Housing material:
    Painted steel
  • Max. RPM:
    650 rpm