ContactUsLinkComponent
Polypropylene and PVC Pressure Regulators
PVC Pressure Regulator Valve, 1/2" NPT(F)
Catalog # MFLX01350-82
undefined
PVC Pressure Regulator Valve, 1/2" NPT(F)
Catalog # MFLX01350-82
Specifications
  • Connection Type:
    NPT(F)
  • Description:
    PVC Pressure Regulator Valve, 1/2" NPT(F)
  • Wetted materials:
    PVC
  • Warranty:
    1 year
  • Pk:
    1
  • Inlet port:
    1/2" NPT(F)
  • Outlet port:
    1/2" NPT(F)
Specifications
About this item
Easily control your outlet pressure.
  • Choose between PVC or polypropylene (PP) construction with Viton® diaphragm
Valves feature a diaphragm made of Viton®. Use to control your outlet pressure between 5 and 50 psig; up to 150 psig inlet. Ideal for your liquid applications.
  • Connection Type:
    NPT(F)
  • Description:
    PVC Pressure Regulator Valve, 1/2" NPT(F)
  • Environmentally Preferable:
  • Wetted materials:
    PVC
  • Warranty:
    1 year
  • Pk:
    1
  • Inlet port:
    1/2" NPT(F)
  • Outlet port:
    1/2" NPT(F)