ContactUsLinkComponent
Masterflex® Multi-Turn PTFE Needle Valves, Avantor®
Masterflex® Multi-Turn PTFE Needle Valves, Avantor®
Certificates
undefined
Masterflex® Multi-Turn PTFE Needle Valves, Avantor®
Specifications
  • Model:
  • 06393-71
  • 06393-68
  • 06393-80
  • 06393-66
  • 06393-82
  • 06393-92
  • 06393-90
  • 06393-94
  • 06393-96
  • 06393-60
  • 06393-61
  • 06393-70
  • Int. Ø tubing:
  • 0,375"
  • 0,375"
  • 0,375"
  • 0,375"
  • Orifice:
  • 3,18 mm (0,125")
  • 3,18 mm (0,125")
  • 3,18 mm (0,125")
  • 3,18 mm (0,125")
  • 3,18 mm (0,125")
  • 3,18 mm (0,125")
  • 3,18 mm (0,125")
  • 3,18 mm (0,125")
  • 3,18 mm (0,125")
  • 3,18 mm (0,125")
  • 3,18 mm (0,125")
  • 3,18 mm (0,125")
  • Tubing ext.Ø:
  • 0,25"
  • 0,25"
  • 0,25"
  • 0,25"
  • Wetted materials:
  • Aluminum, PTFE, CTFE
  • Aluminum, PTFE, CTFE
  • Stainless Steel, PTFE, CTFE
  • Aluminum, PTFE, CTFE
  • Stainless Steel, PTFE, CTFE
  • Stainless Steel, PTFE, CTFE
  • Stainless Steel, PTFE, CTFE
  • Stainless Steel, PTFE, CTFE
  • Stainless Steel, PTFE, CTFE
  • Aluminum, PTFE, CTFE
  • Aluminum, PTFE, CTFE
  • Aluminum, PTFE, CTFE
  • Warranty:
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year