ContactUsLinkComponent
Stopcocks with Flow Indicator, PVC
undefined
Stopcocks with Flow Indicator, PVC
  • Connection type:
  • NPT(F)
  • NPT(M)
  • NPT(M) x Barbed
  • Barbed
  • Inlet port:
  • 1/4" NPT(F)
  • 1/4" NPT(M)
  • 1/4" NPT(M)
  • 1/4" to 1/2" Barb
  • Outlet port:
  • 1/4" NPT(F)
  • 1/4" NPT(M)
  • 1/4" Barb
  • 1/4" to 1/2" Barb
  • Int. Ø tubing:
  • 0,5 - 0,625"
  • 0,25 - 0,5"
  • Max. pressure:
  • 150 psi
  • 150 psi
  • 150 psi
  • 150 psi
  • Max. temperature:
  • 48 °C (120 °F)
  • 48 °C (120 °F)
  • 48 °C (120 °F)
  • 48 °C (120 °F)
  • Port size:
  • 0,25 NPT(F)
  • Orifice:
  • 6,35 mm (0,25")
  • 6,35 mm (0,25")
  • 6,35 mm (0,25")
  • 6,35 to 12,7 mm (0,25 - 0,5")
  • Warranty:
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • Wetted materials:
  • PVC
  • PVC
  • PVC
  • PVC
Frequently Bought Together