ContactUsLinkComponent
Hand-Operated Piston Dispensing Drum Pump, Polypropylene
Hand-Operated Piston Dispensing Drum Pump, Polypropylene
Certificates
undefined
Hand-Operated Piston Dispensing Drum Pump, Polypropylene
Specifications
  • Max. temperature:
  • 82 °C (180 °F)
  • 82 °C (180 °F)
  • Max. viscosity:
  • 4000 SSU
  • 4000 SSU
  • Outlet connection:
  • Spout
  • Spout
  • Tube ext.Ø, inlet:
  • 1,27 cm (0,5")
  • 1,27 cm (0,5")
  • Tube ext.Ø, outlet:
  • 1,27 cm (0,5")
  • 1,27 cm (0,5")
  • Tube length, inlet:
  • 81 cm (32")
  • 81 cm (32")
  • Wetted materials:
  • Polypropylene, Polyethylene
  • Polypropylene, Polyethylene
  • Warranty:
  • 1 year
  • 6 Months