ContactUsLinkComponent
Gast Direct-Drive NEMA Type 56 C-Face Motors
Gast Direct-Drive NEMA Type 56 C-Face Motors
Certificates
undefined
Gast Direct-Drive NEMA Type 56 C-Face Motors
Specifications
  • Air exhaust connection:
  • 0,25 NPT(F)
  • 0,25 NPT(F)
  • Air consumption:
  • 132,52 m³/h (78 ft.³/min)
  • 50,97 m³/h (30 ft.³/min)
  • Air inlet connection:
  • 0,25 NPT(F)
  • 0,25 NPT(F)
  • Max. air inlet pressure:
  • 0,4564625 bar (100 psig)
  • 0,4047875 bar (100 psig)
  • Min. motor RPM:
  • 300 rpm
  • 350 rpm
  • Shaft centerline:
  • 8,26 cm (3,25")
  • 8,26 cm (3,25")
  • Shaft Ø:
  • 1,59 cm (0,625")
  • 1,59 cm (0,625")
  • Warranty:
  • 1 year
  • 1 year