Order Entry
Austria
ContactUsLinkComponent
Masterflex® I/P® Variable-Speed Air-Powered Drive, Avantor®
Masterflex® I/P® Variable-Speed Air-Powered Drive, 100 to 650 rpm
  MFLX07589-30
CE Compliant
undefined
Masterflex® I/P® Variable-Speed Air-Powered Drive, 100 to 650 rpm
  MFLX07589-30
CE Compliant

 

  • Maximum Flow Rate:
    17000 ml/min
  • Control type:
    Analog Variable Speed
  • Pump series:
    I/P
  • Description:
    I/P variable-speed air-powered drive.
  • Drive height:
    229 mm (9")
  • Drive IP rating:
    IP 34
  • Drive length:
    282 mm (11,125")
  • Drive width:
    216 mm (8,5")
  • Min. RPM:
    100 rpm
  • Motor hp:
    3/4
  • Open-head sensor:
    No
  • Reversible motor:
    No
  • Speed control:
    ±10%
  • Type of pump heads accepted:
    Standard, Easy-Load, High-Performance
  • Motor:
    560 W
  • Speed:
    Variable
  • Warranty:
    1 year
  • Pk:
    1
  • Housing material:
    Painted steel
  • Max. RPM:
    650 rpm
  • No. of pump heads accepted:
    2

 

 

Pump safely in hazardous environments.

  • Flow rate 0.6 to 13 LPM I/P tubing—flow rate depends on drive rpm, pump head, and tubing size
  • Easily adjust motor speed by using included regulator
  • Effortlessly connect to your existing air supply or compressor
  • The 3/4-hp air-powered motor starts smooth, runs cool, and requires little maintenance
  • ATEX Zone 2 approved; rated to EEXII 3 G c IIC T6; NEC Class 1, Division 2, Groups A, B, C, D, T6

Use this variable-speed air-powered drive when electrical power may be unsafe or impractical. For safe operation, carefully ground to protect from static electricity.

 : Includes regulator with 5-µm air filter and 1/4'' NPT(F) connection, automatic lubricator, 0 to 100 psi pressure gauge, and muffler.
  • Maximum Flow Rate:
    17000 ml/min
  • Control type:
    Analog Variable Speed
  • Pump series:
    I/P
  • Description:
    I/P variable-speed air-powered drive.
  • Environmentally Preferable:
  • Drive height:
    229 mm (9")
  • Drive IP rating:
    IP 34
  • Drive length:
    282 mm (11,125")
  • Drive width:
    216 mm (8,5")
  • Min. RPM:
    100 rpm
  • Motor hp:
    3/4
  • Open-head sensor:
    No
  • Reversible motor:
    No
  • Speed control:
    ±10%
  • Type of pump heads accepted:
    Standard, Easy-Load, High-Performance
  • Motor:
    560 W
  • Speed:
    Variable
  • Warranty:
    1 year
  • Pk:
    1
  • Housing material:
    Painted steel
  • Max. RPM:
    650 rpm
  • No. of pump heads accepted:
    2