ContactUsLinkComponent
Masterflex® I/P® Easy-Load® Pump Heads for Precision Tubing, Avantor®
Masterflex® I/P® Easy-Load® Pump Heads for Precision Tubing, Avantor®
Certificates
undefined
Masterflex® I/P® Easy-Load® Pump Heads for Precision Tubing, Avantor®
Specifications
  • Model:
  • 77602-00
  • 77602-80
  • 77602-30
  • 77602-10
  • 77601-60
  • Gear ratio:
  • 1:1
  • 1:1
  • 1:1
  • 1:1
  • 1:1
  • No. of channels:
  • 1
  • 1
  • 1
  • 1
  • 1
  • Occlusion:
  • Adjustable
  • Adjustable
  • Adjustable
  • Adjustable
  • Adjustable
  • Open-head sensor:
  • No
  • No
  • Yes
  • No
  • No
  • Optimum RPM range:
  • 0,1 - 650 rpm
  • 0,1 - 650 rpm
  • 0,1 - 650 rpm
  • 0,1 - 650 rpm
  • 0,1 - 650 rpm
  • Pump series:
  • I/P
  • I/P
  • I/P
  • I/P
  • I/P
  • Suction lift:
  • 7,6 m (25 ft.)
  • 7,6 m (25 ft.)
  • 7,6 m (25 ft.)
  • 7,6 m (25 ft.)
  • 7,6 m (25 ft.)
  • Tubing retention:
  • Automatic
  • Automatic
  • Automatic
  • Automatic
  • Automatic
  • Tubing sizes accepted:
  • I/P® 26, I/P 73, I/P 82
  • I/P® 26, I/P 73, I/P 82
  • I/P® 26, I/P 73, I/P 82
  • I/P® 26, I/P 73, I/P 82
  • I/P® 26, I/P 73, I/P 82
  • Volume per revolution with I/P 26 tubing:
  • 6,2 ml
  • 6,2 ml
  • 6,2 ml
  • 6,2 ml
  • 6,2 ml
  • Volume per revolution with I/P 73 tubing:
  • 12,3 ml
  • 12,3 ml
  • 12,3 ml
  • 12,3 ml
  • 12,3 ml
  • Volume per revolution with I/P 82 tubing:
  • 20 ml
  • 20 ml
  • 20 ml
  • 20 ml
  • 20 ml
  • Warranty:
  • 2 years
  • 2 years
  • 2 years
  • 2 years
  • 2 years