ContactUsLinkComponent
Masterflex® Adapter Fittings, Sanitary Clamp to Female Threaded, Straight, Avantor®
Masterflex® Fitting, Polypropylene, Straight, Sanitary Clamp to Thread, 3/4" Mini x 1/2" NPT(F)
Catalog # MFLX31200-13
CE certificate
undefined
Masterflex® Fitting, Polypropylene, Straight, Sanitary Clamp to Thread, 3/4" Mini x 1/2" NPT(F)
Catalog # MFLX31200-13
CE certificate
Specifications
  • Connection Type:
    Sanitary to Thread
  • Flow pattern:
    Straight
  • Sanitary clamp size (Port 1):
    ¾"
  • Thread size NPT(F) (port 2):
    ½"
  • Description:
    Fitting, Polypropylene, Straight, Sanitary Clamp to Thread, ¾" x ½" NPT(F)
  • Seq:
    04
  • Pk:
    1
  • Fitting material:
    Polypropylene
  • Sanitary gasket size:
    ¾"
Specifications
About this item
Maintain sanitary conditions.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Meet USP Class VI for pharmaceutical use and FDA, USDA, and 3A standards for food contact
  • All fittings are compatible with Tri-Clamp® connections
  • Excellent chemical resistance; low leachable characteristics
  • Connection Type: 3/4" mini to 1/2" NPT(F)
These fittings are ideal for use with the Masterflex L/S tubing pumps.
  • Connection Type:
    Sanitary to Thread
  • Flow pattern:
    Straight
  • Sanitary clamp size (Port 1):
    ¾"
  • Thread size NPT(F) (port 2):
    ½"
  • Description:
    Fitting, Polypropylene, Straight, Sanitary Clamp to Thread, ¾" x ½" NPT(F)
  • Seq:
    04
  • Environmentally Preferable:
  • Pk:
    1
  • Fitting material:
    Polypropylene
  • Sanitary gasket size:
    ¾"