ContactUsLinkComponent
Masterflex® Ismatec® Minicartridge Pump Heads for Masterflex® L/S® Drives, Avantor®
Masterflex® Ismatec® Minicartridge Pump Heads for Masterflex® L/S® Drives, Avantor®
Certificates
undefined
Masterflex® Ismatec® Minicartridge Pump Heads for Masterflex® L/S® Drives, Avantor®
Specifications
  • Model:
  • 07623-30
  • 07623-10
  • 07623-00
  • Max. flow rate per channel:
  • 52 ml/min
  • 156 ml/min
  • 195 ml/min
  • Min. flow rate per channel:
  • 0,003 ml/min
  • 0,002 ml/min
  • 0,003 ml/min
  • No. of channels:
  • 4
  • 8
  • 8
  • Occlusion:
  • Adjustable
  • Adjustable
  • Adjustable
  • Open-head sensor:
  • No
  • No
  • No
  • Optimum RPM range:
  • 1 - 600 rpm
  • 1 - 600 rpm
  • 1 - 600 rpm
  • Pump series:
  • L/S
  • L/S
  • L/S
  • Tubing retention:
  • Stopped Tubing or Manual
  • Stopped Tubing or Manual
  • Stopped Tubing or Manual
  • Tubing sizes accepted:
  • Three-stop microbore
  • Three-stop microbore
  • Three-stop microbore
  • Warranty:
  • 2 years
  • 1 year
  • 1 year