ContactUsLinkComponent
Masterflex® Normally Open Low-Flow Compact Switches, Avantor®
Masterflex® Normally Open Low-Flow Compact Switches, Avantor®
Certificates
undefined
Masterflex® Normally Open Low-Flow Compact Switches, Avantor®
Specifications
  • Model:
  • 32778-34
  • 32778-33
  • 32778-36
  • 32778-32
  • 32778-31
  • 32778-35
  • 32778-30
  • Min. flow rate:
  • 2,5 ft.³/min
  • 5 ft.³/min
  • 1 ft.³/min
  • 25 ft.³/min
  • Max. operating temperature:
  • 100 °C (212 °F)
  • 100 °C (212 °F)
  • 100 °C (212 °F)
  • 100 °C (212 °F)
  • 100 °C (212 °F)
  • 100 °C (212 °F)
  • 100 °C (212 °F)
  • Wetted materials:
  • Noryl®, 316 stainless steel, and epoxy
  • Noryl®, 316 stainless steel, and epoxy
  • Noryl®, 316 stainless steel, and epoxy
  • Noryl®, 316 stainless steel, and epoxy
  • Noryl®, 316 stainless steel, and epoxy
  • Noryl®, 316 stainless steel, and epoxy
  • Noryl®, 316 stainless steel, and epoxy
  • Height:
  • 1869"
  • 1869"
  • 1869"
  • 1869"
  • 1869"
  • 1869"
  • 1869"
  • Warranty:
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year