ContactUsLinkComponent
Masterflex® Normally Open Flow Switch for Low Pressure-Drop Liquids, Avantor®
Masterflex® Normally Open Flow Switch for Low Pressure-Drop Liquids, Avantor®
Certificates
undefined
Masterflex® Normally Open Flow Switch for Low Pressure-Drop Liquids, Avantor®
Specifications
  • Max. pressure:
  • 200 psi
  • 200 psi
  • 200 psi
  • 200 psi
  • Max. operating temperature:
  • 100 °C (212 °F)
  • 100 °C (212 °F)
  • 100 °C (212 °F)
  • 100 °C (212 °F)
  • Wetted materials:
  • PP, Ryton® PPS, Viton®, and 316 SS
  • PP, Ryton® PPS, Viton®, and 316 SS
  • PP, Ryton® PPS, Viton®, and 316 SS
  • PP, Ryton® PPS, Viton®, and 316 SS
  • Connection:
  • 1/4" male quick connect terminals
  • 1/4" male quick connect terminals
  • 1/4" male quick connect terminals
  • 1/4" male quick connect terminals
  • Power:
  • 20 VA VAC
  • 20 VA VAC
  • 20 VA VAC
  • 20 VA VAC
  • Warranty:
  • 1 year
  • 1 year
  • 1 year
  • 1 year