ContactUsLinkComponent
Blue-White® Direct-Read Variable-Area Flowmeters for Water, Panel-Mount
Blue-White® Direct-Read Variable-Area Flowmeters for Water, Panel-Mount
Certificates
undefined
Blue-White® Direct-Read Variable-Area Flowmeters for Water, Panel-Mount
Specifications
  • Model:
  • 32471-00
  • 32471-02
  • 32471-04
  • 32471-03
  • 32471-01
  • Accuracy:
  • ±5% full-scale
  • ±5% full-scale
  • ±5% full-scale
  • ±5% full-scale
  • ±5% full-scale
  • Float material:
  • PVC
  • 316 Stainless Steel
  • 316 Stainless Steel
  • 316 Stainless Steel
  • 316 Stainless Steel
  • Max. operating temperature:
  • 115,5 °C (240 °F)
  • 115,5 °C (240 °F)
  • 115,5 °C (240 °F)
  • 115,5 °C (240 °F)
  • 115,5 °C (240 °F)
  • Max. pressure:
  • 150 psi
  • 150 psi
  • 150 psi
  • 150 psi
  • 150 psi
  • Warranty:
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year