ContactUsLinkComponent
CPC® Caps for MPC Quick-Disconnect Fittings
CPC® Caps for MPC Quick-Disconnect Fittings
Certificates
undefined
CPC® Caps for MPC Quick-Disconnect Fittings
Certificates
Specifications
  • Max. operating pressure:
  • 60 psi
  • 60 psi
  • Max. temperature:
  • 148 °C (300 °F)
  • 148 °C (300 °F)
  • Max. vacuum:
  • 634 mm Hg (25" Hg)
  • 634 mm Hg (25" Hg)
  • Min. temperature:
  • –40 °C (–40 °F)
  • –40 °C (–40 °F)
  • Sterilisation:
  • Ethylene oxide (ETO) only
  • Ethylene oxide (ETO) only
  • Warranty:
  • None