ContactUsLinkComponent
CPC® Miniature Quick-Disconnect Fittings, Threaded Bodies
CPC® Miniature Quick-Disconnect Fittings, Threaded Bodies
Certificates
undefined
CPC® Miniature Quick-Disconnect Fittings, Threaded Bodies
Specifications
  • Length:
  • 2,1844 cm
  • 2,1844 cm
  • Max. temperature:
  • 82 °C (180 °F)
  • 82 °C (180 °F)
  • Min. temperature:
  • –40 °C (–40 °F)
  • –40 °C (–40 °F)
  • O-Ring material:
  • Buna N
  • Buna N
  • Sterilisation:
  • Autoclave or ethylene oxide (ETO)
  • Autoclave or ethylene oxide (ETO)
  • Valve material:
  • None
  • Acetal
  • Valve spring material:
  • 316 SS
  • Warranty:
  • 3 Months
  • 1 year